Skip to Content
Merck
All Photos(3)

Key Documents

MABF39-I

Sigma-Aldrich

Anti-DC-STAMP Antibody, clone 1A2

clone 1A2, from mouse

Synonym(s):

Dendritic cell-specific transmembrane protein, DC-STAMP, Dendrocyte-expressed seven transmembrane protein, FIND, hDC-STAMP, IL-four-induced protein, Transmembrane 7 superfamily member 4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.45

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1A2, monoclonal

species reactivity

mouse, human

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

General description

Dendritic cell-specific transmembrane protein (UniProt Q9H295; also known as DC-STAMP, Dendrocyte-expressed seven transmembrane protein, FIND, hDC-STAMP, IL-four-induced protein, Transmembrane 7 superfamily member 4) is encoded by the DCSTAMP (also known as TM7SF4) gene (Gene ID 81501) in human. DC-STAMP is a six-transmembrane protein essential for cell-to-cell fusion to form multinucleated osteoclasts (OCs) during osteoclastogenesis. DC-STAMP expression is upregulated among osteoclast precursor (OCP) cells upon exposure to OC-promoting cytokines, such as receptor activator of nuclear factor-κB (NF-κB) ligand (RANKL), and Dcstamp-knockout (KO) mice have few multinucleated TRAP+ OCs and increased bone mass. On the other hand, DC-STAMP overexpression in transgenic (Tg) mice causes accelerated cell-to-cell fusion during OCP differentiation and enhanced bone resorption. DC-STAMP is a six-trasmembrane (a.a. 35-55, 58-78. 98-118, 210-230, 293-313, 377-397) protein, having both its N- and C-terminal ends exposed intracellularly (a.a. 1-34, 398-470). The C-terminal cytoplasmic tail of DC-STAMP contains an immunoreceptor tyrosine-based inhibitory motif or ITIM sequence (407-SFYPSV-412) that, when phosphorylated on the tyrosine residue, recruits SHP-1. DC-STAMP neutralizing antibody blocks OC formation in vitro and abolishes cellular DC-STAMP and SHP-1 tyrosine phosphorylation.

Specificity

Clone 1A2 recognizes an epitope conserved between human and murine DC-STAMP in the third extracellular domain.

Immunogen

Epitope: The third extracellular domain.
KLH-conjugated linear peptide corresponding a sequence from the third extracellular domain of human DC-STAMP.

Application

Anti-DC-STAMP Antibody, clone 1A2 is an antibody against DC-STAMP for use in Western Blotting, Flow Cytometry, Immunocytochemistry, Immunoprecipitation.
Research Category
Inflammation & Immunology
Research Sub Category
Osteobiology
Western Blotting Analysis: 4.0 µg/mL from a representative lot detected DC-STAMP in 10 µg of thymus lysates from wild-type, but not Dcstamp-knockout mice.
Western Blotting Analysis: 1.0 µg/mL from a representative lot detected DC-STAMP in thymus and spleen lysates from wild-type mice, and greatly reduced DC-STAMP in thymus and spleen lysates from Dcstamp-knockout mice (Courtesy of Grace Chiu, Ph.D., University of Rochester Medical Center, NY, USA).
Western Blotting Analysis: A representative lot detected the ~106 kDa dimeric and the ~53 kDa monomeric DC-STAMP band by Western blotting under non-denatured and denatured condition, respectively, following DC-STAMP immunoprecipitation using murine RAW 264.7 macrophage lysate (Mensah, K.A., et al. (2010). J. Cell. Physiol. 223(1):76-83).
Flow Cytometry Analysis: A representative lot detected DC-STAMP-positive lymphoctes and monocytes in purified human PBMCs (Chiu, Y.H., et al. (2012). J. Bone Miner. Res. 27(1):79-92).
Flow Cytometry Analysis: A representative lot detected DC-STAMP surface expression on murine RAW 264.7 macrophages and murine bone marrow-derived CD11b+ monocytes. DC-STAMP is expressed on osteoclast precursor (OCP) cells as a dimer, which is efficiently detected by flow cytometry using clone 1A2 (Mensah, K.A., et al. (2010). J. Cell. Physiol. 223(1):76-83).
Function Analysis: A representative lot inhibited RANKL & M-CSF treatment-induced osteoclasts (OC) formation in human PBMCs & monocytes cultures (Chiu, Y.H., et al. (2012). J. Bone Miner. Res. 27(1):79-92).
Function Analysis: A representative lot inhibited RANKL treatment-induced osteoclasts (OC) formation in RAW 264.7 and murine bone marrow macrophages cultures (Mensah, K.A., et al. (2010). J. Cell. Physiol. 223(1):76-83).
Immunohistochemistry Analysis: A representative lot detected DC-STAMP expressionon in multinucleated ‘osteoclast-like’ giant cells in human giant cell tumor of bone (Chiu, Y.H., et al. (2012). J. Bone Miner. Res. 27(1):79-92).
Immunocytochemistry Analysis: A representative lot detected DC-STAMP-positive human PBMCs using 10% NBF-fixed, paraffin-embedded cell preparation (Chiu, Y.H., et al. (2012). J. Bone Miner. Res. 27(1):79-92).
Immunocytochemistry Analysis: A representative lot detected differential DC-STAMP intracellular localizations by fluorescent immunocytochemistry staining of 4% paraformaldehyde-fixed, 0.1% saponin-permeabilized murine bone marrow macrophages at different time points during osteoclastogenesis upon RANKL treatment (Mensah, K.A., et al. (2010). J. Cell. Physiol. 223(1):76-83).
Immunoprecipitation Analysis: A representative lot immunoprecipitated DC-STAMP from the lysates of human monocytes (Chiu, Y.H., et al. (2012). J. Bone Miner. Res. 27(1):79-92).
Immunoprecipitation Analysis: A representative lot immunoprecipitated DC-STAMP from the membrane extracts of murine RAW 264.7 macrophages (Mensah, K.A., et al. (2010). J. Cell. Physiol. 223(1):76-83).

Quality

Evaluated by Western Blotting in mouse thymus lysate.

Western Blotting Analysis: 4.0 µg/mL of this antibody detected DC-STAMP in 10 µg of mouse thymus lysate.

Target description

~58 kDa observed. Target band size appears larger than the calculated molecular weights of 53.39 kDa (human) and 53.88 kDa (mouse) due to glycosylation. Uncharacterized band(s) may appear in some lysates.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2aκ antibody in PBS without preservatives.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jarred M Whitlock et al.
Nature communications, 14(1), 616-616 (2023-02-05)
Multinucleated osteoclasts, essential for skeletal remodeling in health and disease, are formed by the fusion of osteoclast precursors, where each fusion event raises their bone-resorbing activity. Here we show that the nuclear RNA chaperone, La protein has an additional function
Yuji Inagaki et al.
Journal of clinical medicine, 10(3) (2021-01-28)
Gan-Lu-Yin (GLY), a traditional Chinese herbal medicine, shows therapeutic effects on periodontitis, but that mechanism is not well known. This study aims to clarify the precise mechanism by investigating the inhibitory effects of GLY extracts on osteoclastogenesis in vitro and
Ya-Hui Chiu et al.
Journal of cellular physiology, 232(9), 2538-2549 (2016-10-11)
DC-STAMP is a multi-pass transmembrane protein essential for cell-cell fusion between osteoclast precursors during osteoclast (OC) development. DC-STAMP-/- mice have mild osteopetrosis and form mononuclear cells with limited resorption capacity. The identification of an Immunoreceptor Tyrosine-based Inhibitory Motif (ITIM) on
Mariana S P Ribeiro et al.
The Journal of biological chemistry, 299(12), 105379-105379 (2023-10-24)
Osteoclasts are specialized cells responsible for bone resorption, a highly energy-demanding process. Focus on osteoclast metabolism could be a key for the treatment of osteolytic diseases including osteoporosis. In this context, AMP-activated protein kinase α1 (AMPKα1), an energy sensor highly
Mariana Pena Ribeiro Soares et al.
Cell biology international, 43(5), 466-475 (2019-02-15)
Reactive oxygen species (ROS) are produced by NADPH oxidase (NOX), an enzyme that reduces oxygen by using NADPH as a substrate. Apocynin (APO) is a catechol that is used as a NOX inhibitor, and N-acetyl-cysteine ​​(NAC) can reduce intracellular ROS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service