Skip to Content
Merck
All Photos(1)

Key Documents

Z4250

Sigma-Aldrich

Zymosan A from Saccharomyces cerevisiae

for inducing inflamatory response

Synonym(s):

Yeast Cell Wall

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.61

biological source

Saccharomyces cerevisiae

form

powder with small lumps

technique(s)

flow cytometry: suitable
tissue culture: suitable

color

off-white to light brown

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

  • in flow cytometry
  • to study its effect on head kidney leukocytes from rainbow trout (Oncorhynchus mykiss)

Zymosan A from Saccharomyces cerevisiae has been used:
  • as a positive control to determine the activation of complement cascade
  • for respiratory burst (RB) assay
  • to assess its effects on performance, immunity, heat shock protein and apoptosis related gene expressions of dairy cows under heat stress (HS)
  • a stimulating agent in Krebs-Ringer phosphate (KRP) medium
  • to induce thermal hyperalgesia in mice

Biochem/physiol Actions

Zymosan is prepared from yeast cell wall and consists of protein-carbohydrate complexes. It is used to induce experimental sterile inflammation. In macrophages, zymosan-induced responses include the induction of proinflammatory cytokines, arachidonate mobilization, protein phosphorylation, and inositol phosphate formation. Zymosan A also raises cyclin D2 levels suggesting a role for the latter in macrophage activation besides proliferation. It potentiates acute liver damage after galactosamine injection suggesting that certain types of nonparenchymal cells other than Kupffer cells are involved in zymosan action. Zymosan A, a 13 beta glucan contributes to the development of organic dust-induced respiratory disease.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The G2A receptor controls polarization of macrophage by determining their localization within the inflamed tissue
Kern K, et al.
Frontiers in Immunology, 9, 2261-2261 (2018)
Impact of copper nanoparticles on porcine neutrophils: ultrasensitive characterization factor combining chemiluminescence information and USEtox assessment model
Pu Y, et al.
Materials Today Communications, 11, 68-75 (2017)
Protective effects of zymosan on heat stress-induced immunosuppression and apoptosis in dairy cows and peripheral blood mononuclear cells
Sun Y, et al.
Cell Stress & Chaperones, 23(5), 1069-1078 (2018)
ACUTE INFLAMMATION AND RECOVERY IN RATS AFTER INTRATRACHEAL INSTILLATION OF A 1?3-
Young SH, et al.
Journal of Toxicology and Environmental Health. Part A, 64(4), 311-325 (2001)
PAMP induced expression of immune relevant genes in head kidney leukocytes of rainbow trout (Oncorhynchus mykiss)
Chettri JK, et al.
Developmental and Comparative Immunology, 35(4), 476-482 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service