Skip to Content
Merck
All Photos(1)

Documents

MAB6020

Sigma-Aldrich

Anti-Cytochrome P450 11β-Hydroxylase Antibody, clone 11B2-1A4

culture supernatant, clone 11B2-1A4, Chemicon®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

culture supernatant

antibody product type

primary antibodies

clone

11B2-1A4, monoclonal

species reactivity

rat

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

Specificity

Rat p450 11beta-Hydroxylase

Immunogen

Aa 272-285 (KNVYRELAE GRQQS-C)

Application

Anti-Cytochrome P450 11β-Hydroxylase Antibody, clone 11B2-1A4 is an antibody against Cytochrome P450 11β-Hydroxylase for use in IH(P) & WB.
Research Category
Metabolism
Research Sub Category
Enzymes & Biochemistry
Western blot: 1:300

Immunohistochemistry (S.T.F. on paraffin embedded tissue required, traditional formalin fixation or paraformaldehyde fixation is not recommended. Microwave or pressure cooker antigen retrieval; blocking with a solution of 0.5% SDS, 2% milk and 5% NGS in Tris-HCL pH 7.6. Primary antibody is diluted in PBS or TBS typically with no SDS or NGS: 1:50-200, usually overnight refrigerated. After washing 4X with Tris buffer pH 7.6, secondary goat anti-mouse secondary (AP124B) is incubated (1:500-1:2000) diluted in a solution of 2% milk, 5% NGS, Tris-HCL pH 7.6 and 0.05% tween 20. Detection is via streptavidin HRP/DAB. Recommended counter stain is Gill′s Haematoxylin.

Optimal working dilutions must be determined by end user.

STF fixative is available from streck laboratories, www.streck.com).

Physical form

Supernatant. Liquid in 0.1% sodium azide.

Storage and Stability

Maintain refrigerated at 2-8°C in undiluted aliquots for up to 6 months from date of receipt.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chuanming Xu et al.
American journal of physiology. Cell physiology, 311(4), C686-C695 (2016-08-19)
(Pro)renin receptor (PRR) is predominantly expressed in the collecting duct (CD) with unclear functional implication. It is not known whether CD PRR is regulated by high potassium (HK). Here, we aimed to investigate the effect of HK on PRR expression
Peter I Imoesi et al.
Frontiers in molecular neuroscience, 12, 259-259 (2019-11-19)
Retinoic acid (RA) is the active metabolite of vitamin A but is also used as a medication, primarily for acne in which the treatment regime lasts several months. A number of studies have indicated that treatment with RA over this
Chuanming Xu et al.
American journal of physiology. Renal physiology, 313(3), F641-F656 (2016-07-22)
(Pro)renin receptor (PRR) is highly expressed in the distal nephron, but it has an unclear functional implication. The present study was conducted to explore a potential role of renal PRR during high K+ (HK) loading. In normal Sprague-Dawley rats, a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service