Skip to Content
Merck
All Photos(1)

Documents

MAB3400

Sigma-Aldrich

Anti-Vimentin Antibody, clone V9

clone V9, Chemicon®, from mouse

Synonym(s):

Vimentin antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

V9, monoclonal

species reactivity

monkey, feline, pig, bovine, rat, human, chicken, canine

should not react with

mouse

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... VIM(7431)

General description

Vimentin is the major subunit protein of the intermediate filaments of mesenchymal cells. It is believed to be involved with the intracellular transport of proteins between the nucleus and plasma membrane. Vimentin has been implicated to be involved in the rate of steroid synthesis via its role as a storage network for steroidogenic cholesterol containing lipid droplets. Immunohistochemical staining for Vimentin is characteristic of sarcomas (of neural, muscle and fibroblast origin) compared to carcinomas which are generally negative. Melanomas, lymphomas and vascular tumors may all stain for Vimentin.

Specificity

Recognizes a 57-60 kDa protein. Shows no cross reactivity with other closely related intermediate filament proteins including desmin and GFAP. In tissue sections, this reagent shows positive reaction on connective tissue, endothelial cells, vascular smooth muscle cells and astrocytes. In tissue culture, typical intermediate filament profiles are seen in fibroblasts and other cell types, which contain vimentin

Immunogen

Purified vimentin from pig eye lens

Application

Anti-Vimentin Antibody, clone V9 is an antibody against Vimentin for use in IC, IH, IH(P) & WB.
Immunoblotting:
A dilution of 1:1000 was used in a previous lot.

Immunohistochemistry:
5-10 μg/mL was used on a previous lot; can be used on formalin fixed paraffin embedded tissues with traditional high temperature, citrate buffer treatments. Prolonged formalin fixation can reduce antivbody reactivity.

Immunocytochemistry:
5-10 μg/mL of a previous lot was used in IC.

Optimal working dilutions must be determined by end user.

Quality

Evaluated by Western Blot on HeLa lysates.

Western Blot:
1:1000 dilution of this antibody detected VIMENTIN on 10 μg of HeLa lysates

Target description

57-60 kDa

Linkage

Replaces: MAB3410

Physical form

Format: Purified
Purified mouse monoclonal IgG1 presented in 0.02 M Phosphate buffer, 0.25 M NaCl with 0.1% sodium azide.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Noelia López-Montero et al.
Autophagy, 12(10), 1886-1901 (2016-08-04)
Xenophagy has been studied in epithelial cells infected with Salmonella enterica serovar Typhimurium (S. Typhimurium). Distinct autophagy receptors target this pathogen to degradation after interacting with ubiquitin on the surface of cytosolic bacteria, and the phagophore- and autophagosome-associated protein MAP1LC3/LC3.
E Mato et al.
Journal of molecular endocrinology, 52(3), 289-300 (2014-03-20)
Tumor malignancy is associated with the epithelial-mesenchymal transition (EMT) process and resistance to chemotherapy. However, little is known about the relationship between the EMT and the multidrug-resistance gene in thyroid tumor progression. We investigated whether the expression of the ABCG2/BCRP
Muhammad Umar Cheema et al.
PloS one, 9(7), e101258-e101258 (2014-07-08)
Prolonged elevations of plasma aldosterone levels are associated with renal pathogenesis. We hypothesized that renal distress could be imposed by an augmented aldosterone-induced protein turnover challenging cellular protein degradation systems of the renal tubular cells. Cellular accumulation of specific protein
Dolores D Mruk et al.
Biology of reproduction, 80(3), 590-601 (2008-12-17)
During spermatogenesis, leptotene spermatocytes residing in the basal compartment of the seminiferous epithelium must traverse the blood-testis barrier (BTB) to gain entry into the adluminal compartment for further development. At the same time, these as well as other germ cell
Shih-Pei Lin et al.
PloS one, 7(11), e49605-e49605 (2012-11-28)
Hypoxia and serum depletion are common features of solid tumors that occur upon antiangiogenesis, irradiation and chemotherapy across a wide variety of malignancies. Here we show that tumor cells expressing CD133, a marker for colorectal cancer initiating or stem cells

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service