Skip to Content
Merck
All Photos(1)

Documents

MAB201P

Sigma-Aldrich

Mouse Anti-Rabbit light chain, HRP conjugate Antibody

Chemicon®, from mouse

Synonym(s):

HRP Anti-Rabbit, Light Chain Detection Antibody, Rabbit Light Chain Detection

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

peroxidase conjugate

antibody form

purified antibody

antibody product type

secondary antibodies

clone

monoclonal

species reactivity

rabbit

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

shipped in

wet ice

target post-translational modification

unmodified

General description

Antibody molecules typically comprise two immunoglobulin light chains covalently bound to a pair of heavy chains. Immunoglobulin light chains occur in two types, designated by the Greek letters kappa and lambda. Kappa and gamma light chains are approximately 250 amino acids in length with an average mass of about 25 kDa. The ratio of kappa to lambda found in the immunoglobulin population varies by species.

Specificity

Minimal cross-reaction with bovine, goat, Armenian hamster, horse, human, mouse, rat, and sheep.
The antibody reacts strongly with native primary antibodies primarily with kappa light chains. It is not suitable for detecting lambda light chains. The antibody does not react with the heavy chain of rabbit IgG. The antibody has been tested by ELISA and adsorbed to ensure minimal cross-reaction with bovine, goat, Armenian hamster, horse, human, mouse, rat, and sheep immunoglobulins.

Immunogen

Epitope: Kappa light chain
Prepared from rabbit IgG light chain.

Application

Detect Rabbit light chain using this Mouse anti-Rabbit light chain, HRP conjugate Antibody validated for use in ELISA, WB, IC, IH.
Research Category
Secondary & Control Antibodies
Research Sub Category
Fragment Specific Secondary Antibodies
Western Blotting: 1:5,000 -1:100,000 with chromogenic substrates 1:10,000 -1:200,000 with ECL substrates
ELISA: 1:5,000 -1:100,000
Immunohistochemistry: 1:500 - 1:5,000
Immunocytochemistry: 1:500 - 1:5,000

Optimal working dilutions must be determined by the end user.

Target description

25 kDa

Physical form

Purified from antiserum by chromatographic procedures. Lyophilized in 0.01M Sodium Phosphate, 0.25M NaCl, pH 7.6. 15 mg/mL BSA as stabilizer.
Serum

Storage and Stability

Maintain lyophilized product at 2°-8°C for up to 12 months. After reconstitution the product is stable for six weeks at 2°-8°C in the dark as an undiluted liquid. Reconstitute vial with of distilled water (addition of 0.5 ml water will yield a final concentration of 1 mg/mL). For extended storage after reconstitution, add an equal volume of glycerol to make a final concentration of 50% glycerol followed by storage at -20°C in undiluted aliquots for up to 12 months. Please note the concentration of protein (and buffer salts) will decrease to one-half of the original after the addition of glycerol. Avoid repeated freeze thaw cycles. Keep away from light.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aneta Dobierzewska et al.
PloS one, 11(6), e0157221-e0157221 (2016-06-11)
Preeclampsia (PE), is a serious pregnancy disorder characterized in the early gestation by shallow trophoblast invasion, impaired placental neo-angiogenesis, placental hypoxia and ischemia, which leads to maternal and fetal morbidity and mortality. Here we hypothesized that angiogenic sphingosine kinase-1 (SPHK1)/sphingosine-1-phosphate
Asunción Muñoz-Silvestre et al.
The American journal of pathology, 190(6), 1188-1210 (2020-03-24)
Despite the enormous efforts made to achieve effective tools that fight against Staphylococcus aureus, the results have not been successful. This failure may be due to the absence of truly representative experimental models. To overcome this deficiency, the present work
María Rodríguez-Muñoz et al.
Translational psychiatry, 7(12), 1291-1291 (2017-12-19)
The activity of certain G protein-coupled receptors (GPCRs) and of glutamate N-Methyl-D-aspartate receptors (NMDARs) is altered in both schizophrenia and depression. Using postmortem prefrontal cortex samples from subjects with schizophrenia or depression, we observed a series of opposite changes in
Nomi Pernicone et al.
Molecules (Basel, Switzerland), 27(3) (2022-02-16)
DNA-damaging chemotherapy agents such as cisplatin have been the first line of treatment for cancer for decades. While chemotherapy can be very effective, its long-term success is often reduced by intrinsic and acquired drug resistance, accompanied by chemotherapy-resistant secondary malignancies.
Jürgen Fritsch et al.
Oncotarget, 7(46), 75774-75789 (2016-10-08)
During apoptosis induction by TNF, the extrinsic and intrinsic apoptosis pathways converge at the lysosomal-mitochondrial interface. Earlier studies showed that the lysosomal aspartic protease Cathepsin D (CtsD) cleaves Bid to tBid, resulting in the amplification of the initial apoptotic cascade

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service