Skip to Content
Merck
All Photos(1)

Key Documents

MAB1581

Sigma-Aldrich

Anti-Chondroitin Sulfate Proteoglycan Antibody, Core Protein Epitope, clone Cat-315

ascites fluid, clone Cat-315, Chemicon®

Synonym(s):

CSPG

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

Cat-315, monoclonal

species reactivity

feline, rat

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgM

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... ACAN(176)

Specificity

Chondroitin sulfate proteoglycan (CSPG), protein core epitope

Immunogen

Epitope: Core Protein Epitope

Application

Immunohistochemistry: 1:2,000 on 4% paraformaldehyde fixed frozen tissue.

Immunocytochemistry: 1:500 on primary cultures of neurons.

Immunoblot: 1:5,000 recognizes a band at 680 kDa.

Immunoprecipitation.

Optimal working dilutions must be determined by the end user.
This Anti-Chondroitin Sulfate Proteoglycan Antibody, Core Protein Epitope, clone Cat-315 is validated for use in IP, WB, IC, IH for the detection of Chondroitin Sulfate Proteoglycan.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paulette A McRae et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 27(20), 5405-5413 (2007-05-18)
An important role for the neural extracellular matrix in modulating cortical activity-dependent synaptic plasticity has been established by a number of recent studies. However, identification of the critical molecular components of the neural matrix that mediate these processes is far
Andrew Nathaniel Stewart et al.
Restorative neurology and neuroscience, 35(4), 395-411 (2017-06-10)
Utilizing genetic overexpression of trophic molecules in cell populations has been a promising strategy to develop cell replacement therapies for spinal cord injury (SCI). Over-expressing the chemokine, stromal derived factor-1 (SDF-1α), which has chemotactic effects on many cells of the
Claudia Loebel et al.
Advanced functional materials, 30(44) (2021-07-03)
Hydrogels are engineered with biochemical and biophysical signals to recreate aspects of the native microenvironment and to control cellular functions such as differentiation and matrix deposition. This deposited matrix accumulates within the pericellular space and likely affects the interactions between
Eleanor Grant et al.
Cerebral cortex (New York, N.Y. : 1991), 26(3), 1336-1348 (2016-01-09)
Corticothalamic projection systems arise from 2 main cortical layers. Layer V neurons project exclusively to higher-order thalamic nuclei, while layer VIa fibers project to both first-order and higher-order thalamic nuclei. During early postnatal development, layer VIa and VIb fibers accumulate
Alicia L Hawthorne et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 30(2), 420-430 (2010-01-15)
Embryonic CNS neurons can migrate from the ventricular zone to their final destination by radial glial-guided locomotion. Another less appreciated mechanism is somal translocation, where the young neuron maintains its primitive ventricular and pial processes, through which the cell body

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service