Skip to Content
Merck
All Photos(1)

Documents

LSKS096

Millipore

Montage SEQ96 Sequencing Reaction Cleanup Kit

Purification of sequencing reactions

Synonym(s):

Multiscreen SEQ

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104923
eCl@ss:
32039006
NACRES:
NA.52

sterility

non-sterile

manufacturer/tradename

Montage®

technique(s)

DNA sequencing: suitable

plate size

96 wells

application(s)

genomic analysis

shipped in

ambient

General description

The Montage SEQ96 sequencing reaction cleanup kit provides the filtration plate and solution necessary to remove contaminating salts and unincorporated dye terminators from DNA sequencing reactions. The vacuum-based size-exclusion separation platform eliminates the need for gel filtration or bind-wash elute columns that require centrifugation. The entire protocol requires very little time to process typically less than 10 minutes. In addition, the optimized filtration area enables reaction miniaturization and reagent reduction, resulting in lower DNA sequencing costs. Montage SEQ96 sequencing reaction clean-up kit consists of 96 well SEQ plate and an injection solution.

Application

Montage SEQ96 Sequencing Reaction Cleanup Kit is suitable for use:

  • in the purification of polymerase chain reaction (PCR) products and to remove unincorporated primers and dye terminators for mutational analysis of Krüppel-like factor 4 (KLF4)
  • in the purification of sequencing reactions
  • in 16S rDNA sequence analysis to remove unincorporated dye terminators from the sequencing reaction

Components

includes 96-well filter plates and injection solution

Physical form

Color Code: Blue

Other Notes

Automation Compatible: No

Legal Information

MONTAGE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Weissella ghanensis sp. nov., isolated from a Ghanaian cocoa fermentation
De Bruyne K
International Journal of Systematic and Evolutionary Microbiology (2008)
KLF4 is a novel candidate tumor suppressor gene in pancreatic ductal carcinoma
Zammarchi F, et al.
The American Journal of Pathology (2011)
Diversity analysis of diazotrophic bacteria associated with the roots of tea (Camellia sinensis (L.) O. Kuntze)
Gulati A, et al.
Journal of microbiology and biotechnology (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service