SC1000
SigmaPrep™ spin column
storage temp.:20-25°C, pkg of 1 kit
Synonym(s):
dna extraction, dna purification, spin columns
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
40161600
NACRES:
NB.22
Recommended Products
packaging
pkg of 1 kit
storage temp.
20-25°C
General description
Benefits:
- Convenient format for both use and storage
- Compatible with both microcentrifuges and vacuum manifolds
- Technical bulletin provides protocol suggestions
Legal Information
SigmaPrep is a trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Jinhee Kim et al.
Biomolecules & therapeutics, 28(3), 282-291 (2020-04-28)
Inhaled solvents such as toluene are of particular concern due to their abuse potential that is easily exposed to the environment. The inhalation of toluene causes various behavioral problems, but, the effect of short-term exposure of toluene on changes in
Anup Shrestha et al.
Scientific reports, 10(1), 11388-11388 (2020-07-11)
Steroid receptor coactivator-3 (SRC-3) regulates the activity of both nuclear hormone receptors and a number of key transcription factors. It is implicated in the regulation of cell proliferation, inflammation and in the progression of several common cancers including breast, colorectal
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service