Skip to Content
Merck
All Photos(1)

Key Documents

LSKPMRN30

Millipore

RNase A, 0.9 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116012
eCl@ss:
32160405
NACRES:
NA.54

form

liquid

Quality Level

packaging

pkg of 0.9 mL

manufacturer/tradename

Montage®

technique(s)

molecular cloning: suitable (BAC prep)
molecular cloning: suitable (plasmid miniprep)

shipped in

ambient

General description

The Montage Plasmid MiniprepHTS 96 Kit is a fast, easy-to-use kit for high-purity plasmid or bacterial artificial chromosome (BAC) minipreps

Application

Research Category
All
Suitable for plasmid & BAC minipreps

Physical form

30mg in 50% glycerol

Storage and Stability

Reagents should be stored at 15 °C to 30 °C.

Legal Information

MONTAGE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service