Skip to Content
Merck

Key Documents

Skip To

R4875

Ribonuclease A from bovine pancreas

Type I-A, powder, ≥60% RNase A basis (SDS-PAGE), ≥50 Kunitz units/mg protein

Synonym(s):

Pancreatic Ribonuclease, RNAsea, RNase A, Ribonucleate 3′-pyrimidinooligonucleotidohydrolase

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
NACRES:
NA.54
UNSPSC Code:
12352204
EC Number:
232-646-6
MDL number:
EC Number:
Specific activity:
≥50 Kunitz units/mg protein
Biological source:
bovine pancreas
Concentration:
≥60% (RNase A, SDS-PAGE)
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

bovine pancreas

Quality Segment

type

Type I-A

form

powder

specific activity

≥50 Kunitz units/mg protein

mol wt

~13,700

concentration

≥60% (RNase A, SDS-PAGE)

technique(s)

cell based assay: suitable

impurities

salt, essentially free

input

general use sample(s)

suitability

suitable for molecular biology

application(s)

diagnostic assay manufacturing

foreign activity

protease, essentially free

storage temp.

−20°C

SMILES string

[nH]1cncc1CC(NC(=O)CCN)C(=O)O

InChI

1S/C9H14N4O3/c10-2-1-8(14)13-7(9(15)16)3-6-4-11-5-12-6/h4-5,7H,1-3,10H2,(H,11,12)(H,13,14)(H,15,16)

InChI key

CQOVPNPJLQNMDC-UHFFFAOYSA-N

General description

RNase A, Ribonuclease A, is an endoribonuclease that cleaves the phosphodiester bonds of single strand RNA after pyrimidine nucleotides. It attacks at the 3′ phosphate end (For example pG-pG-pC-pA-pG will be cleaved to give pG-pG-pCp and A-pG). The highest activity is exhibited with single stranded RNA. RNase A is a single chain polypeptide containing 4 disulfide bridges. In contrast to RNase B, it is not a glycoprotein. Ribonucleases do not hydrolyze DNA, because the DNA lacks 2′-OH groups essential for the formation of cyclic intermediates. RNase A can also hydrolyze RNA from protein samples. RNase A can be inhibited by alkylation of His12 and His119 and activated by potassium and sodium salts. RNAse is inhibited in the presence of heavy metal ions. RNase is also inhibited competitively by DNA.

Application

  • RNase A is used to remove RNA from DNA plasmid and genomic DNA preparations and protein samples.
  • RNase A is also used in RNA sequence analysis and protection assays.
  • RNase A has been used as a tool for computer-aided drug design.
  • RNase A supports the analysis of RNA sequences.
  • RNase A hydrolyze RNA contained in protein samples.
  • Purification of DNA is supported by RNase A.

Biochem/physiol Actions

Ribonuclease A is an endoribonuclease that cleaves single stranded RNA after pyrimidine nucleotides. It attacks at the 3′ phosphate end. Ribonucleases do not hydrolyze DNA, because the DNA lacks 2′-OH groups essential for the formation of cyclic intermediates. RNase can also hydrolyze RNA from protein samples. RNase A can be inhibited by alkylation of His12 and His119 and activated by potassium and sodium salts.

Features and Benefits

Our highly stable Ribonuclease A, RNase A, is suitable for removal of RNA, RNA sequencing, and DNA purification.

Preparation Note

Salt fractionated and chromatographically purified.

Analysis Note

Protein determined by E.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
R5500R5000R4642
description

Type I-A, powder, ≥60% RNase A basis (SDS-PAGE), ≥50 Kunitz units/mg protein

description

Type XII-A, ≥90% (SDS-PAGE), 75-125 Kunitz units/mg protein

description

Type II-A, ≥60% (SDS-PAGE), >= 60 Kunitz units/mg protein

description

(Solution of 50% glycerol, 10mM Tris-HCL pH 8.0)

biological source

bovine pancreas

biological source

bovine pancreas

biological source

bovine pancreas

biological source

bovine pancreas

specific activity

≥50 Kunitz units/mg protein

specific activity

75-125 Kunitz units/mg protein

specific activity

>= 60 Kunitz units/mg protein

specific activity

-

technique(s)

cell based assay: suitable

technique(s)

cell based assay: suitable

technique(s)

cell based assay: suitable

technique(s)

-

suitability

suitable for molecular biology

suitability

suitable for mRNA or total RNA extracted from cells and tissues

suitability

suitable for molecular biology

suitability

suitable for

form

powder

form

lyophilized powder

form

solid

form

(Solution of 50% glycerol, 10mM Tris-HCL pH 8.0)

concentration

≥60% (RNase A, SDS-PAGE)

concentration

-

concentration

≥60% (SDS-PAGE)

concentration

20-40 mg/mL


Still not finding the right product?


Pictograms

Health hazard

Signal Word

Danger

Hazard Codes

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters