LSKMAGLM02
PureProteome Lambda Ig Binder Magnetic Beads
The PureProteome Lambda Ig Binder Magnetic Bead System is a powerful system that helps researchers purify all Immunoglobulins with high specificity.
Synonym(s):
Magnetic Beads for Ig Binding (Lambda), PureProteome Magnetic Beads
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116133
eCl@ss:
32160405
NACRES:
NA.56
Recommended Products
packaging
pkg of 2 mL
manufacturer/tradename
PureProteome
technique(s)
protein purification: suitable (antibody)
particle size
10 μm
capacity
≥2.0 μg/μL, slurry binding capacity (lambda IgG)
shipped in
wet ice
storage temp.
2-8°C
General description
The PureProteome Lambda Ig Binder Magnetic Bead System is a powerful system that helps researchers purify all Immunoglobulins (i.e IgG, IgA, IgM, IgE and IgD) with high specificity. Researchers can also purify/seperate the Fab portion from the Fc portion of Immunoglobulins.
Application
The PureProteome Lambda Ig Binder Magnetic Beads are suitable for antibody and protein purification.
Features and Benefits
- PureProteome™ magnetic beads can be used manually or with automated systems.
- Suitable for low, medium, or high-throughput sample preparation.
- Saves time and costs for researchers to process multiple samples in parallel.
- PureProteome™ magnetic beads possess paramagnetic properties, making them ideal for automation.
Legal Information
PureProteome is a trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class Code
12 - Non Combustible Liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service