Skip to Content
Merck
All Photos(2)

Key Documents

HTH17MAG-14K

Millipore

MILLIPLEX® Human TH17 Magnetic Bead Panel - Immunology Multiplex Assay

Simultaneously analyze multiple Th17 cytokine and chemokine biomarkers with the Th17 Bead-Based Multiplex Assays using the Luminex technology, in human serum, plasma and cell culture samples.

Synonym(s):

Human Th17 cytokine multiplex kit, Luminex® human Th17 cytokine immunoassay, Millipore human Th17 cytokine panel

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.84

Quality Level

species reactivity

human

manufacturer/tradename

Milliplex®

assay range

accuracy: 79-107%
standard curve range: 1-5000 pg/mL
(IL-10)

standard curve range: 10-40,000 pg/mL
(IFNγ)

standard curve range: 12-50,000 pg/mL
(IL-17A & IL-2)

standard curve range: 24-100,000 pg/mL
(IL-17 & IL-4)

standard curve range: 3-10,000 pg/mL
(IL-6 & TNFα)

standard curve range: 336-1,500,000 pg/mL
(IL-23)

standard curve range: 37-150,000 pg/mL
(IL-22 & TNFβ)

standard curve range: 488-2,000,000 pg/mL
(IL-17E/IL-25)

standard curve range: 49-200,000 pg/mL
(IL-28a & IL-31)

standard curve range: 5-20,000 pg/mL
(MIP-3α, IL-1β, IL-12 (p70), IL-15, IL-21 & IL-33)

standard curve range: 6-25,000 pg/mL
(IL-5)

standard curve range: 61-250,000 pg/mL
(GM-CSF & IL-27)

standard curve range: 7-30,000 pg/mL
(IL-13)

standard curve range: 9-35,000 pg/mL
(IL-9)

technique(s)

multiplexing: suitable

detection method

fluorometric (Luminex xMAP)

shipped in

wet ice

General description

CD4 T-helper cells are major players in adaptive immunity. Based on their expression profile of transcription factors and secreted cytokines, these cells are divided into the major subsets Th1, Th2, Th17 and T regulatory (Treg) cells. Th1 cells, induced by IL-12 and enhanced by IFNγ, secrete IFNγ that activates macrophages. Induced by IL-4, Th2 cells primarily produce IL-4, IL-5, IL-13, IL-25 and IL-17E, playing a role in allergy. Treg cells, induced by TGFβ, produce the anti-inflammatory cytokines IL-10 and TGFβ and function to control T-cell responses to prevent autoimmune reactions. Th17 cells, a more recently discovered subset characterized by the production of IL-17, are induced by TGFβ combined with IL-23 and IL-1β in humans, and TGFβ, IL-6 and IL-21 in mice. Activated Th17 cells secrete IL-17A, IL-17F, IL-21, IL-22 and TNFα to promote tissue inflammation.

Th17 cells are involved in the clearance of extracellular bacteria and fungi. They are abundant in the intestinal lamina propria and function as a barrier against invading pathogens. Excessive amounts of Th17 cells have been implicated in the pathogenesis of several autoimmune diseases, including multiple sclerosis, psoriasis, juvenile diabetes, rheumatoid arthritis, Crohn′s disease, and autoimmune uveitis. In addition, Th17 cells play an important role in tumor development, progression, and metastasis, potentially in both promoting and inhibiting tumor growth.

MILLIPLEX® Human TH17 Bead Panel is a 25-plex kit to be used for the simultaneous quantification of the following analytes in serum, plasma or culture supernatant samples: GM-CSF, IFN-γ, IL-1β, IL-2, IL-4, IL-5, IL-6, IL-9, IL-10, IL-12 (p70), IL-13, IL-15, IL-17A, IL-17F, IL-17E/IL-25, IL-21, IL-22, IL-23, IL-27, IL-28A, IL-31, IL-33, MIP-3α/CCL20, TNF-α and TNFβ.

The Luminex® xMAP® platform uses a magnetic bead immunoassay format for ideal speed and sensitivity to quantitate multiple analytes simultaneously, dramatically improving productivity while conserving valuable sample volume.

Panel Type: Cytokines/Chemokines

Specificity

Cross Reactivty
Cross-reactivity between the antibodies and any of the other analytes in this panel is non-detectable or negligible.

Application

  • Analytes: GM-CSF, IFN-γ, IL-1β, IL-2, IL-4, IL-5, IL-6, IL-9, IL-10, IL-12 (p70), IL-13, IL-15, IL-17A, IL-17F, IL-17E/IL-25, IL-21, IL-22, IL-23, IL-27, IL-28A, IL-31, IL-33, MIP-3α/CCL20, TNF-α, TNFβ
  • Recommended Sample type: Serum, plasma, and cell culture samples
  • Recommended Sample dilution: Neat
  • Assay Run Time: Overnight
  • Research Category: Inflammation & Immunology

Features and Benefits

Design your multiplex kit by choosing available analytes within this panel.

Storage and Stability

Recommended storage for kit components is 2 - 8°C.

Other Notes

Sensitivity: Refer to kit protocol for sensitivities of individual biomarkers.

Legal Information

Luminex is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Sens. 1 - STOT RE 2

Target Organs

Respiratory Tract

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Suzanne Cohen et al.
Nature communications, 6, 8327-8327 (2015-09-15)
In response to infections and irritants, the respiratory epithelium releases the alarmin interleukin (IL)-33 to elicit a rapid immune response. However, little is known about the regulation of IL-33 following its release. Here we report that the biological activity of
Marta Czesnikiewicz-Guzik et al.
European heart journal, 40(42), 3459-3470 (2019-09-11)
Inflammation is an important driver of hypertension. Periodontitis is a chronic inflammatory disease, which could provide a mechanism for pro-hypertensive immune activation, but evidence of a causal relationship in humans is scarce. We aimed to investigate the nature of the
Olivier Fogel et al.
The Journal of allergy and clinical immunology, 142(1), 258-268 (2017-09-17)
An interferon signature is involved in the pathogenesis of primary Sjögren syndrome (pSS), but whether the signature is type 1 or type 2 remains controversial. Mouse models and genetic studies suggest the involvement of TH1 and type 2 interferon pathways.
Danielle W Lowe et al.
Pediatric research, 82(1), 55-62 (2017-01-19)
Vitamin D has neuroprotective and immunomodulatory properties, and deficiency is associated with worse stroke outcomes. Little is known about effects of hypoxia-ischemia or hypothermia treatment on vitamin D status in neonates with hypoxic-ischemic encephalopathy (HIE). We hypothesized vitamin D metabolism
Gunita Deksne et al.
Vector borne and zoonotic diseases (Larchmont, N.Y.), 21(1), 1-5 (2020-09-29)
The zoonotic parasite Dirofilaria repens has spread toward north in Europe, and cases of autochthonous dirofilariosis caused by D. repens have emerged in the Baltic countries Estonia, Latvia, and Lithuania. We conducted a review on the emergence of dirofilariosis in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service