Skip to Content
Merck
All Photos(4)

Key Documents

HCYTA-60K

Millipore

MILLIPLEX® Human Cytokine/Chemokine/Growth Factor Panel A - Immunology Multiplex Assay

Synonym(s):

Human cytokine multiplex kit, Luminex® human cytokine immunoassay, Millipore human cytokine panel

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

species reactivity

human

Quality Level

manufacturer/tradename

Milliplex®

technique(s)

multiplexing: suitable

input

cell culture supernatant(s)
plasma
serum

detection method

fluorometric (Luminex® xMAP®)

shipped in

wet ice

storage temp.

2-8°C

General description

Cytokines, chemokines, and growth factors are key mediatorsof immune system functions capable of signaling through autocrine, paracrine,and endocrine mechanisms. Their pleiotropic immunomodulatory propertiesallow these biomolecules to react to diverse stimuli and regulate the immuneresponse, either promoting or inhibiting inflammation.

Application

  • MILLIPLEX® Qualified assays undergo rigorousassay development, verification, and Quality Control testing to achieve optimalperformance.
  • Simultaneously analyze up to 48 cytokine,chemokine, and growth factor biomarkers with bead-based multiplex assays usingLuminex technology, in human serum, plasma and cell culture samples.
  • Available analytes: sCD40L, EGF, Eotaxin, FGF-2,FLT-3L, Fractalkine, G-CSF, GM-CSF, GROα, IFNα2, IFNγ, IL-1α, IL-1β, IL-1RA,IL-2, IL-3, IL-4, IL-5, IL-6, IL-7, IL-8, IL-9, IL-10, IL-12 (p40), IL-12(p70), IL-13, IL-15, IL-17A, IL-17E/IL-25, IL-17F, IL-18, IL-22, IL-27, IP-10,MCP-1, MCP-3, M-CSF, MDC, MIG, MIP-1α, MIP-1β, PDGF-AA, PDGF-AB/BB, RANTES,TGFα, TNFα, TNFβ, VEGF-A

Features and Benefits

  • This panel is fully configurable, with options for custom bead premixing, fixed premixed and bulk premixed panels.
  • Save time and resources by simultaneously analyzing up to 48 analytes of your choice in a single sample
  • Requires a small sample volume (25 µL/well)
  • Choose same day or overnight incubation
  • Reliable and accurate performance with proven lot-to-lot consistency
  • Ready-to-use reagents
  • Comes with everything you need to run your assay
  • Includes a Serum Matrix to mimic the native sample environment in serum and plasma samples
  • Includes high and low Quality Controls

Packaging

96-well plate

Storage and Stability

Recommended storage for kit components is 2 - 8°C.

Legal Information

Luminex is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp

Disclaimer

Unless otherwise stated in our catalogor other company documentation accompanying the product(s), our products areintended for research use only and are not to be used for any other purpose,which includes but is not limited to, unauthorized commercial uses, in vitrodiagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumptionor application to humans or animals.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Sens. 1 - STOT RE 2

Target Organs

Respiratory Tract

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carl A Pierce et al.
JCI insight, 6(9) (2021-04-07)
BACKGROUNDCoronavirus disease 2019 (COVID-19) is more benign in children compared with adults for unknown reasons. This contrasts with other respiratory viruses where disease manifestations are often more severe in children. We hypothesize that a more robust early innate immune response
Yiwei Zhao et al.
Fertility and sterility, 115(4), 1044-1053 (2020-12-05)
To compare the changing peripheral levels of inflammation-related cytokine profile during a 9-day period after blastocyst transfer between women who did and did not conceive. Prospective, observational, and longitudinal study. University-affiliated hospital. Forty-seven women with infertility who were undergoing single
Carl A Pierce et al.
medRxiv : the preprint server for health sciences (2021-02-18)
COVID-19 is more benign in children compared to adults for unknown reasons. This contrasts with viruses such as influenza where disease manifestations are often more severe in children1. We hypothesized that a more robust early innate immune response to SARS-CoV-2
Andrea R Daamen et al.
Frontiers in immunology, 13, 989556-989556 (2022-10-04)
COVID-19 manifests a spectrum of respiratory symptoms, with the more severe often requiring hospitalization. To identify markers for disease progression, we analyzed longitudinal gene expression data from patients with confirmed SARS-CoV-2 infection admitted to the intensive care unit (ICU) for
Shawn J Rice et al.
PloS one, 16(12), e0260988-e0260988 (2021-12-03)
Blood-based biomarkers including systemic inflammation (SI) indicators or circulating factors (cytokines, chemokines, or growth factors) are associated with a poor prognosis for lung cancer patients. Collectively these biomarkers can predict the immune state of a patient. We wanted to define

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service