Skip to Content
Merck
All Photos(1)

Key Documents

FTP65400

Millipore

Gasket, O-Ring, 202.8 X 3.53 Mm, EPDM Peroxyde Cured, Set Of 10

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116500

Pricing and availability is not currently available.

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
P3556P4279P7443
solubility

water: 50 mg/mL, clear, colorless to very faintly yellow

solubility

-

solubility

-

solubility

-

form

powder

form

lyophilized powder

form

solution

form

lyophilized powder

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

biological source

soybean

biological source

egg yolk

biological source

egg yolk

biological source

soybean


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Need A Sample COA?

This is a sample Certificate of Analysis (COA) and may not represent a recently manufactured lot of this specific product.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service