Skip to Content
Merck
All Photos(1)

Key Documents

CP74

Sigma-Aldrich

Anti-Vinculin Mouse mAb (V284 or VLN01)

liquid, Calbiochem®, from mouse

Synonym(s):

Anti-Vinculin antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

V284, monoclonal
VLN01, monoclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

human

manufacturer/tradename

Calbiochem®

storage condition

do not freeze

isotype

IgG1

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... VCL(7414)

General description

Protein G purified mouse monoclonal antibody generated by immunizing BALB/c mice with the specified immunogen and fusing splenocytes with mouse myeloma SP2/0 cells. Recognizes the ~130 (vinculin) and ~150 kDa (meta-vinculin) proteins.
Recognizes the ~130 (vinculin) and ~150 kDa (meta-vinculin) proteins in skin tissue.
This Anti-Vinculin Mouse mAb (V284 or VLN01) is validated for use in Immunoblotting, Immunoprecipitation, Paraffin Sections for the detection of Vinculin.

Immunogen

Human
semi-purified, human vinculin

Application


Immunoblotting (1 g/ml, chemiluminescence)
Immunoprecipitation (2 g/ml lysate)
Paraffin Sections (4-8 g/ml, heat pre-treatment required)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In 10 mM PBS, 0.2% BSA, pH 7.4.

Analysis Note

Negative Control
Normal mouse serum
Positive Control
Skin cells or tissue

Other Notes

Palovuori, R., and Eskelinen, S. 2000 Eur. J. Cell Biol.79, 961.
Bakolitsa, C., et al. 1999 Cell99, 603.
Rudiger, M., et al. 1998 Bioessays20, 733
Tidball, J.G., et al. 1999 J. Biol. Chem.274, 33155
Goldmann, W.H., et al. 1996 J. Muscle Res. Cell Motil.17, 1.
Asijee, G.M., et al. 1990 Eur. J. Biochemistry189, 131
Otto, J.J. 1990 Cell Motil. Cytoskeleton16, 1.
This antibody is useful for studies of the role of vinculin in platelet activation. Vinculin (Ab-1) exhibits membrane and cytoplasmic staining when used in immunohistochemistry. For staining of formalin-fixed paraffin sections, boil in 10 mM citrate buffer, pH 6.0, for 10-20 min, followed by cooling at room temperature for 20 min. Antibody should be titrated for optimal results in individual systems.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Harikrishnareddy Paluvai et al.
Molecular oncology, 12(12), 2165-2181 (2018-10-14)
Expression of the class IIa HDACs is frequently altered in different human cancers. In mouse models these transcriptional repressors can trigger transformation, acting as bona fide oncogenes. Whether class IIa HDACs also exhibit transforming activities in human cells is currently
Job de Lange et al.
American journal of cancer research, 2(5), 492-507 (2012-09-08)
The p53 tumor suppressor pathway is inactivated in cancer either via direct mutation or via deregulation of upstream regulators or downstream effectors. P53 mutations are rare in uveal melanoma. Here we investigated the role of the p53 inhibitor Hdmx in
A Freije et al.
Oncogene, 31(50), 5180-5192 (2012-02-22)
Human epidermis is continuously exposed to environmental mutagenic hazard and is the most frequent target of human cancer. How the epidermis coordinates proliferation with differentiation to maintain homeostasis, even in hyperproliferative conditions, is unclear. For instance, overactivation of the proto-oncogene
Rebekah L Mokry et al.
mBio, 11(6) (2020-12-17)
Nitric oxide is a versatile and critical effector molecule that can modulate many cellular functions. Although recognized as a regulator of infections, the inhibitory mechanism of nitric oxide against human cytomegalovirus (HCMV) replication remains elusive. We demonstrate that nitric oxide
Steve Seltzsam et al.
Translational oncology, 12(3), 417-425 (2018-12-17)
Human papillomavirus (HPV) associated squamous cell carcinomas of the head and neck region (HPV+ HNSCCs) harbor diverging biological features as compared to classical noxa-induced (HPV-) HNSCC. One striking difference between subtypes is that the tumor suppressor gene TP53 is usually

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service