Skip to Content
Merck
All Photos(1)

Key Documents

BM-1RYSJ1

Millipore

Auto2D® Glycine Type Reagent Kit

reagent kit for the Auto2D® 2-D Electrophoresis Device

Synonym(s):

2D Electrophoresis, 2DE, 2DE Reagent, Glycine, Tris-Glycine, Tris-Glycine Reagent Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.25

product line

Auto2D®

technique(s)

SDS-PAGE: suitable
western blot: suitable

storage temp.

2-8°C

General description

Glycine Type Reagent Kit is the basic reagent kit used for the Auto2D® 2-D Electrophoresis Device.

Application

  • Differential proteomic analysis
  • Post-translational modification
  • Protein Isoforms
  • Western Blotting

Components

  • Rehydration Solution Kit
  • DTT Kit
  • Glycine Type Equilibration Buffer
  • Anode + Buffer
  • Glycine Type Cathode - Buffer

Legal Information

Auto2D is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Dam. 1 - Met. Corr. 1 - Repr. 2 - Skin Irrit. 2

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service