Skip to Content
Merck
All Photos(1)

Key Documents

AP193P

Sigma-Aldrich

Donkey Anti-Guinea Pig IgG Antibody, HRP conjugate, Species Adsorbed

1 mg/mL (Reconstituted), Chemicon®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46

biological source

donkey

Quality Level

conjugate

peroxidase conjugate

antibody form

F(ab′)2 fragment of affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

guinea pig

manufacturer/tradename

Chemicon®

concentration

1 mg/mL (Reconstituted)

technique(s)

ELISA: suitable
western blot: suitable

shipped in

wet ice

target post-translational modification

unmodified

Application

Donkey anti-Guinea Pig IgG Antibody, HRP conjugate, Species Adsorbed detects level of Donkey Guinea Pig IgG & has been published & validated for use in ELISA & WB.
ELISA: 1:5,000-1:100,000.Western blots: 1:5,000-1:100,000.

Immunohistochemistry: 1:500-1:5,000.

Optimal working dilutions must be determined by end user.

Physical form

Lyophilized. Buffer = 0.01 M Sodium Phosphate, 0.25 M NaCl, pH 7.6 with 15 mg/mL BSA. Contains no preservative.

RECONSTITUTION:

Reconstitute with sterile distilled water to match the volume indicated on the vial label. Centrifuge product if it is not completely clear after standing for 1-2 hours at room temperature.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kentaro Mizuta et al.
American journal of respiratory cell and molecular biology, 39(3), 296-304 (2008-04-12)
Gamma-aminobutyric acid (GABA) is the major inhibitory neurotransmitter in the mammalian central nervous system and exerts its actions via both ionotropic (GABA(A)) and metabotropic (GABA(B)) receptors. The GABA(B) receptor is a dimer composed of R1 and R2 components and classically
Daniel Stadlbauer et al.
Nature communications, 13(1), 7864-7864 (2022-12-22)
Contemporary influenza A H3N2 viruses circulating since 2016 have acquired a glycosylation site in the neuraminidase in close proximity to the enzymatic active site. Here, we investigate if this S245N glycosylation site, as a result of antigenic evolution, can impact
Marwin Groener et al.
Kidney360, 1(6), 480-490 (2020-04-16)
Clathrin-mediated endocytosis (CME) plays a fundamental role in podocyte health. Genetic ablation of genes implicated in CME has been shown to cause severe proteinuria and foot process effacement in mice. However, little is known about the cargo of clathrin-coated vesicles
Rupali Gund et al.
Autophagy, 19(1), 296-305 (2022-06-03)
Alopecia areata (AA) involves an aberrant immune attack on the hair follicle (HF), which leads to hair loss. Previous genetic data from our lab pointed to a connection between macroautophagy/autophagy and AA pathogenesis, and GWAS identified STX17, CLEC16A and BCL2L11/BIM
Anders Madsen et al.
Immunity, 53(4), 852-863 (2020-09-26)
Influenza B virus (IBV) infections can cause severe disease in children and the elderly. Commonly used antivirals have lower clinical effectiveness against IBV compared to influenza A viruses (IAV). Neuraminidase (NA), the second major surface protein on the influenza virus

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service