Skip to Content
Merck
All Photos(1)

Key Documents

AP112B

Sigma-Aldrich

Goat Anti-Human IgG Antibody, biotin conjugate

Chemicon®, from goat

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46

biological source

goat

Quality Level

conjugate

biotin conjugate

antibody form

affinity purified immunoglobulin

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

shipped in

wet ice

target post-translational modification

unmodified

General description

IgG is a monomeric immunoglobulin, consists of two heavy chains (gamma chains) and two light chains. Each molecule has two antigen binding sites. IgG is the most abundant immunoglobulin. It is approximately equally distributed in blood and in interstitial fluid, constituting 75% of serum immunoglobulins in humans. This is the only isotype that can pass through the human placenta, providing protection to the fetus in its first weeks of life before its own immune system has developed. There are 4 subclasses: IgG1 (66%), IgG2 (23%), IgG3 (7%) and IgG4 (4%).

Specificity

The antibody reacts with the heavy chains of human IgG and with the light chains common to most human immunoglobulins.

Immunogen

Prepared from purified human IgG.

Application

EIA: 1:20,000-1:400,000 using enzyme-streptavidin conjugate.
Western blots: 1:20,000-1:400,000 using enzyme-streptavidin conjugate.
Immunohistochemistry: 1:500-1:5,000 using enzyme-streptavidin conjugate.
Flow cytometry: 1:200-1:1,000.
Fluorescence Immunohisto/cytochemistry: 1:200-1:1,1000.

Optimal working dilutions must be determined by the end user.
Research Category
Secondary & Control Antibodies
Research Sub Category
Whole Immunoglobulin Secondary Antibodies
This Goat anti-Human IgG Antibody, biotin conjugate is validated for use in ELISA, WB for the detection of Human IgG.

Physical form

ImmunoAffinity Purified
Purified by immunoaffinity chromatography using antigen coupled to agarose beads. Lyophilized. Buffer is 0.01 M Sodium Phosphate, 0.25 M NaCl, pH 7.6 with 15 mg/mL BSA, and 0.05% sodium azide.

Storage and Stability

Maintain lyophilized product at 2°-8°C for up to 12 months from date of receipt. After reconstitution the product is stable for six weeks at 2°-8°C as an undiluted liquid. Reconstitute vial with 2 mL of distilled water. For extended storage after reconstitution, add an equal volume of glycerol to make a final concentration of 50% glycerol followed by storage at -20°C in undiluted aliquots for up to 12 months. Please note the concentration of antibody (and buffer salts) will decrease to one-half of the original after the addition of glycerol and final working dilutions must be adjusted accordingly. Avoid repeated freeze thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stefania Rosito et al.
Journal of cellular and molecular medicine, 22(2), 1236-1246 (2017-10-22)
Neuromyelitis optica (NMO) is an autoimmune demyelinating disease of the central nervous system (CNS) caused by autoantibodies (NMO-IgG) against the water channel aquaporin-4 (AQP4). Though AQP4 is also expressed outside the CNS, for example in skeletal muscle, patients with NMO
Francesco Pisani et al.
Cells, 8(2) (2019-02-06)
The CNS plasma-membrane water channel aquaporin-4 (AQP4) is expressed as two major isoforms able to aggregate into supramolecular assemblies known as 'orthogonal arrays of particles' (OAPs). OAP subnanometric features are largely unknown mainly because a method for the expression, isolation
Qun Zhou et al.
mAbs, 12(1), 1814583-1814583 (2020-09-08)
Antibodies mediate effector functions through Fcγ receptor (FcγR) interactions and complement activation, causing cytokine release, degranulation, phagocytosis, and cell death. They are often undesired for development of therapeutic antibodies where only antigen binding or neutralization would be ideal. Effector elimination
Benjamin Geary et al.
Frontiers in immunology, 14, 1290585-1290585 (2023-12-14)
MZB1 is an endoplasmic reticulum residential protein preferentially expressed in plasma cells, marginal zone and B1 B cells. Recent studies on murine B cells show that it interacts with the tail piece of IgM and IgA heavy chain and promotes
Praveen K Amancha et al.
Journal of immunology (Baltimore, Md. : 1950), 191(12), 6060-6070 (2013-11-15)
The programmed cell death-1 (PD-1)/programmed cell death ligand-1 pathway has been shown to limit cell-mediated effector functions during chronic viral infections impeding clearance of pathogens. As a strategy to reverse this exhaustion and increase T cell polyfunctionality, PD-1 ligands were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service