Skip to Content
Merck
All Photos(2)

Documents

ABD116

Sigma-Aldrich

Anti-Human Oct-4 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

POU domain,class 5, transcription factor 1, NF-A3, Octamer-binding protein 3, Oct-3, Octamer-binding protein 4, Oct-4, Octamer-binding transcription factor 3, OTF-3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.75

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

chimpanzee (based on 100% sequence homology), goat (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... POU5F1(5460)

General description

Octamer-4 (Oct-4), a member of the POU family of transcription factors, has been demonstrated to be vital for the formation of self-renewing pluripotent stem cells. During embryogenesis expression of Oct-4 is limited to pluripotent cells of the inner cell mass (ICM) that contribute to the formation of all fetal cell types. This relationship between Oct-4 and pluripotency has seen this transcription factor emerge as a marker of pluripotent stem cells. Undifferentiated human and murine pluripotent Embryonic Stem (ES) and Embryonic Carcinoma (EC) cells express Oct-4. Additionally, murine Embryonic Germ (EG) cells are also known to express Oct-4. Following stem cell differentiation, the level of Oct-4 expression decreases. Oct-4 expression by human EG cells is not known.

Specificity

Reacts with human Oct-4 only, and detects the Oct-4A isoform. (Note: Please order Catalog No. MAB4419 for use with mouse embryonic stem cells).
The clone reacts strongly with specific nuclear binding to human ES cells and cross-reacts weakly with mouse ES cells.

Immunogen

Epitope: unknown
KLH-EDC-conjugated linear peptide corresponding to the unknown of Human Oct-4.

Application

Immunocytochemistry Analysis: A 1:1000 dilution from a representative lot detected human Oct-4 in 10 Āµg of H9 human ES cells.
Research Category
Stem Cell Research

Stem Cell Research
Research Sub Category
Neural Stem Cells

Developmental Signaling
This Anti-Human Oct-4 Antibody is validated for use in Western Blotting and Immunocytochemistry for the detection of Human Oct-4.

Quality

Evaluated by Western Blotting in H9 human ES cell lysate and mouse ES cells lysate.

Western Blotting Analysis: A 0.25 Āµg/mL dilution of this antibody detected human Oct-4 in 10 Āµg of H9 human ES cell lysate but not in 10 Āµg of mouse ES cells lysate.

Target description

~40 kDa observed

Physical form

Antigen Affinity Purified
Purified rabbit Polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8Ā°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product? Ā 

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a productā€™s label following the words ā€˜Lotā€™ or ā€˜Batchā€™.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isabel Y Buchsbaum et al.
EMBO reports, 21(5), e48204-e48204 (2020-03-25)
During embryonic development, excitatory projection neurons migrate in the cerebral cortex giving rise to organised layers. Periventricular heterotopia (PH) is a group of aetiologically heterogeneous disorders in which a subpopulation of newborn projection neurons fails to initiate their radial migration

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service