Skip to Content
Merck
All Photos(2)

Key Documents

MABD24

Sigma-Aldrich

Anti-NANOG Antibody, clone 7F7.1

clone 7F7.1, from mouse

Synonym(s):

Homeobox protein NANOG, Homeobox transcription factor Nanog, hNanog

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

7F7.1, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... NANOG(79923)

General description

NANOG (Homeobox protein NANOG) is a member of the Nanog homeobox family of DNA-binding proteins. It is expressed in embryonic stem cells and confers pluripotency on these cells. Once embryonic stem cells become differentiated, NANOG expression is suppressed. NANOG is involved in the Hedgehog/Gli1 signaling pathway which has been implicated in the development and growth of various types of tumors.

Immunogen

GST-tagged recombinant protein corresponding to human NANOG.

Application

Anti-NANOG Antibody, clone 7F7.1 is an antibody against NANOG for use in Western Blotting, ICC.
Immunocytochemistry Analysis: A 1:500 dilution from a representative lot detected NANOG in H9 human embryonic stem cells.
Research Category
Stem Cell Research
Research Sub Category
Transcription Factors

Quality

Evaluated by Western Blot in H9 human embryonic stem cell lysate.

Western Blot Analysis: 0.5 µg/mL of this antibody detected NANOG on 10 µg of H9 human embryonic stem cell lysate.

Target description

~39 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
H9 human embryonic stem cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ambrin Fatima et al.
Stem cell research, 44, 101739-101739 (2020-03-04)
Incontinentia pigmenti (IP) is an X-linked dominant neuroectodermal dysplasia caused by loss-of-function mutations in the IKBKG gene. Using CRISPR/Cas9 technology, we generated an IKBKG knock-out iPSC line (KICRi002-A-1) on a 46,XY background. The iPSC line showed a normal karyotype, expressed
Yifan Zhou et al.
Molecular autism, 7, 42-42 (2016-10-08)
Fragile X syndrome (FXS), a common cause of intellectual disability and autism, results from the expansion of a CGG-repeat tract in the 5' untranslated region of the FMR1 gene to >200 repeats. Such expanded alleles, known as full mutation (FM)
Ambrin Fatima et al.
Stem cell research, 44, 101758-101758 (2020-03-24)
The role of Neurochondrin (NCDN) in humans is not well understood. Mice with a conditional Ncdn knock-out show epileptic seizures, depressive-like behaviours and impaired spatial learning. Using CRISPR/Cas9, we generated a Neurochondrin deficient human iPSC line KICRi002-A-3 carrying a homozygous
Victor J T Lin et al.
Scientific reports, 7(1), 5005-5005 (2017-07-12)
Despite their well-known function in maintaining normal cell physiology, how inorganic elements are relevant to cellular pluripotency and differentiation in human pluripotent stem cells (hPSCs) has yet to be systematically explored. Using total reflection X-ray fluorescence (TXRF) spectrometry and inductively
Alexandra Moran et al.
Oncology reports, 44(1), 393-403 (2020-07-07)
Chondrosarcoma is the second most common primary malignant bone tumor and is resistant to chemotherapy and radiation. Inadequate treatment response and poor prognosis requires novel therapeutic approaches. Proline‑rich polypeptide‑1 (PRP‑1), synthesized by brain neurosecretory cells, has demonstrated antitumor properties in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service