Skip to Content
Merck
All Photos(1)

Key Documents

AB986

Sigma-Aldrich

Anti-β Galactosidase Antibody, bacterial

serum, Chemicon®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

E. coli, vertebrates

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... GLB1(2720)

General description

The previously assigned protein identifier P16279 has been merged into P16278. Full details can be found on the UniProt database.

Specificity

Specific for Beta galactosidase from E coli by IEP. May cross react with Beta galactosidase from other species.

Immunogen

Beta galactosidase from E-coli.
Epitope: bacterial

Application

Anti-β Galactosidase Antibody, bacterial detects level of β Galactosidase & has been published & validated for use in ELISA, IP, WB, IH.
Immunoblotting: 1:500-1:2,000

Immunoprecipitation

Immunohistochemistry (Zhang, 2002)

ELISA: 1:2,000-1:10,000 when tested against 1 μg of immunogen by a standard sandwich ELISA.

Optimal working dilutions must be determined by the end user.
Research Category
Metabolism
Research Sub Category
Enzymes & Biochemistry

Physical form

Rabbit antisera prepared by delipidation and defibrination. Lyophilized. Reconstitute with 2 mL of sterile deionized water. Contains 0.02M Potassium Phosphate, 0.15M NaCl and 0.01% sodium azide after reconstitution.

Storage and Stability

Maintain lyophilized material at 2-8°C for up to 12 months. After reconstitution maintain in undiluted aliquots at -20°C for up to 6 months. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas Visone et al.
Biology of reproduction, 81(3), 571-579 (2009-05-22)
In murine testes, only Sertoli cells express the cathepsin L (Ctsl) gene, and this expression is restricted to stages V-VIII of the cycle. Our previous transgenic analysis of Tg (-2065/+977) demonstrated that this expression is regulated by a approximately 2-kb
Mesodiencephalic dopaminergic neuronal differentiation does not involve GLI2A-mediated SHH-signaling and is under the direct influence of canonical WNT signaling.
Mesman, S; von Oerthel, L; Smidt, MP
Testing null
J Sedý et al.
Developmental dynamics : an official publication of the American Association of Anatomists, 235(4), 1081-1089 (2006-02-24)
ER81, a member of the ETS family of transcription factors, is involved in processes of specification of neuronal identity, control of sensory-motor connectivity, and differentiation of muscle spindles. Spindles either degenerate or are abnormal in mutant mice lacking ER81. We
Dushani L Palliyaguru et al.
Free radical biology & medicine, 101, 116-128 (2016-11-05)
Small molecules of plant origin offer presumptively safe opportunities to prevent carcinogenesis, mutagenesis and other forms of toxicity in humans. However, the mechanisms of action of such plant-based agents remain largely unknown. In recent years the stress responsive transcription factor
Susanne Walter et al.
Acta neuropathologica, 137(2), 239-257 (2018-11-15)
Brain accumulation and aggregation of amyloid-β (Aβ) peptides is a critical step in the pathogenesis of Alzheimer's disease (AD). Full-length Aβ peptides (mainly Aβ1-40 and Aβ1-42) are produced through sequential proteolytic cleavage of the amyloid precursor protein (APP) by β-

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service