Skip to Content
Merck
All Photos(1)

Key Documents

AB1211

Sigma-Aldrich

Anti-β Galactosidase Antibody

Chemicon®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

E. coli, vertebrates

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... GLB1(2720)

Specificity

Recognizes Beta-Galactosidase (E. coli). Monospecific by IEP. Some cross reactivity with Beta Galactosidase from other species may occur.

Immunogen

Beta-Galactosidase (E. coli).

Application

Anti-β Galactosidase Antibody is an antibody against β Galactosidase for use in ELISA, Immunohistochemistry, Immunoprecipitation, Western Blotting.
ELISA: 1:10,000-1:50,000 when assayed against 1 μg of Beta Galactosidase in a standard sandwich ELISA.

Immunoblotting: 1:2000-1:10,000 on beta-galactosidase fusion proteins.

Immunohistochemistry: 1:1,000-1:5000 on tissue fixed with 4% paraformaldehyde.

Immunoprecipitation

Optimal working dilutions must be determined by end user.
Research Category
Epitope Tags & General Use
Research Sub Category
Epitope Tags

Physical form

Ammonium sulfate precipitation and DEAE-cellulose chromatography
Format: Purified
Immunoglobulin Purified by delipidation, salt fractionation and ion-exchange chromatography. Liquid in 0.02M Potassium Phosphate buffer, 0.15M Sodium Chloride, with 0.01% sodium azide.

Storage and Stability

Maintain for 2 years at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Proteins tagged with beta galactosidase

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Su Jung Hwang et al.
Cell death and differentiation (2022-01-03)
Ninjurin1 (Ninj1), an adhesion molecule, regulates macrophage function in hyaloid regression, multiple sclerosis, and atherosclerosis. However, its biological relevance and the mechanism underlying its function in vascular network integrity have not been studied. In this study, we investigated the role
Alejandra E Arbetman et al.
Journal of virology, 79(24), 15238-15245 (2005-11-25)
Preexisting humoral immunity to adeno-associated virus (AAV) vectors may limit their clinical utility in gene delivery. We describe a novel caprine AAV (AAV-Go.1) capsid with unique biological properties. AAV-Go.1 capsid was cloned from goat-derived adenovirus preparations. Surprisingly, AAV-Go.1 capsid was
Tobias B Kurth et al.
Arthritis and rheumatism, 63(5), 1289-1300 (2011-05-04)
We previously reported that human synovium contains cells that, after culture expansion, display properties of mesenchymal stem cells (MSCs). The objective of this study was to identify MSCs in native synovium in vivo. To identify stem cells in the synovium
S C Nagel et al.
Endocrinology, 142(11), 4721-4728 (2001-10-19)
We have developed a transgenic mouse that functions as a reporter of ER activity, termed ER action indicator (ERIN), by incorporating a transgene with an estrogen-responsive promoter (three copies of the vitellogenin estrogen response element with a minimal thymidine kinase
Toshiyuki Sakamaki et al.
Molecular and cellular biology, 26(14), 5449-5469 (2006-07-01)
The cyclin D1 gene encodes a regulatory subunit of the holoenzyme that phosphorylates and inactivates the pRb tumor suppressor to promote nuclear DNA synthesis. cyclin D1 is overexpressed in human breast cancers and is sufficient for the development of murine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service