Skip to Content
Merck
All Photos(1)

Key Documents

AB9272

Sigma-Aldrich

Anti-Synaptophysin Antibody

Chemicon®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, rat

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SYP(6855)

Specificity

Synaptophysin

Immunogen

Synthetic peptide from human synaptophysin.

Application

Anti-Synaptophysin Antibody detects level of Synaptophysin & has been published & validated for use in WB, IH(P).
Research Category
Neuroscience
Research Sub Category
Synapse & Synaptic Biology
Western blot

Immunohistochemistry on formalin fixed, paraffin embedded tissue: 1:300 for 10 minutes at room temperature. Staining of formalin-fixed tissues requires boiling of tissues in 10 mM citrate buffer, pH 6.0 for 10 minutes followed by cooling at room temperature for 20 minutes.

Optimal working dilutions must be determined by the end user.

STAINING PATTERN: Cytoplasmic

Linkage

Replaces: 04-1019

Physical form

Affinity purified immunoglobulin. Liquid in 10 mM PBS, pH 7.4 containing BSA and 0.1% sodium azide.

Storage and Stability

Maintain at 2-8°C for up to 6 months after date of receipt.

Analysis Note

Control
POSITIVE CONTROL: Pancreas or Pheochromocytoma

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Goran Tomic et al.
Cell stem cell, 23(3), 436-443 (2018-08-14)
The intestinal epithelium is largely maintained by self-renewing stem cells but with apparently committed progenitors also contributing, particularly following tissue damage. However, the mechanism of, and requirement for, progenitor plasticity in mediating pathological response remain unknown. Here we show that phosphorylation
Rachel Comba et al.
F1000Research, 4, 396-396 (2016-03-01)
Improvement on spatial tasks is observed during a late, postnatal developmental period (PND18 - PND24).  The purpose of the current work was 1) to determine whether the emergence of spatial-behavioral function was based on the ability to generate appropriate behavioral
Yongfang Chen et al.
Environmental toxicology, 31(12), 1720-1730 (2015-07-29)
The objective of this study is to understand the impairment of learning and memory in mouse after chronic nitrite exposure. The animal model of nitrite exposure in mouse was created with the daily intubation of nitrite in adult healthy male
Stuart D Portbury et al.
PloS one, 12(8), e0183683-e0183683 (2017-08-25)
Traumatic brain Injury (TBI) is a significant cause of death and long-term disability for which there are currently no effective pharmacological treatment options. In this study then, we utilized a mouse model of TBI to assess the therapeutic potential of
Active and passive immunization strategies based on the SDPM1 peptide demonstrate pre-clinical efficacy in the APPswePSEN1dE9 mouse model for Alzheimer's disease.
Camboni, M; Wang, CM; Miranda, C; Yoon, JH; Xu, R; Zygmunt, D; Kaspar, BK; Martin, PT
Neurobiology of Disease null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service