Key Documents
Z719668
Sorenson low binding standard tips
maximum volume 1000 μL, pack of 1000 ea (Bag), non-sterile
About This Item
UNSPSC Code:
41121600
NACRES:
NB.01
volume:
1000 μL
sterility:
non-sterile
feature:
barrier: no
packaging:
pack of 1000 ea (Bag)
case of 10000 ea (10 bags of 100 tips)
manufacturer/tradename:
Sorenson 10190T
material
colorless
polypropylene
sterility
non-sterile
feature
barrier: no
packaging
pack of 1000 ea (Bag)
case of 10000 ea (10 bags of 100 tips)
manufacturer/tradename
Sorenson 10190T
maximum volume
1000 μL
volume
1000 μL
Related Categories
General description
Tips feature a new surface technology designed for pipetting substances which tend to stick to the walls of pipette tips or for pipetting expensive reagents. Universal fit for research-grade pipettors.
Legal Information
Sorenson is a trademark of Sorenson BioScience, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)