Skip to Content
Merck
All Photos(1)

Key Documents

AB3637

Sigma-Aldrich

Anti-DEDAF Antibody

Chemicon®, from rabbit

Synonym(s):

Anti-AAP1, Anti-APAP-1, Anti-DEDAF, Anti-YEAF1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, rat, mouse

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... RYBP(23429)

Specificity

Apoptosis is related to many diseases and induced by a family of cell death receptors and their ligands. Cell death signals are transduced by death domain (DD), death effector domain (DED), and caspase recruitment domain (CARD) containing molecules. DEDAF (DED Associated Factor) is expressed in multiple tissues and cell lines. DEDAF is identical to the transcriptional repressor RYBP. DEDAF interacts with FADD and augments the formation of CD95/FADD/Caspase-8 complexes at the cell membrane. DEDAF also interacts with DEDD (DED-containing DNA binding) protein in the nucleus, which indicates its involvement in the regulation of both cytoplasmic and nuclear events of apoptosis.

Immunogen

Rabbit anti-DEDAF polyclonal antibody was raised against a synthetic peptide corresponding to amino acids 215 to 228 of human-DEDAF. This sequence is identical to the mouse sequence.

Application

Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
This Anti-DEDAF Antibody is validated for use in WB for the detection of DEDAF.
Western blot: 0.5-1 μg/mL.

A549 or HepG2 cell lysate can be used as a positive control and a 32 kDa band should be detected.

Optimal working dilutions must be determined by end user.

Physical form

Format: Purified
Purified IgG by immunoaffinity chromatography. Liquid in PBS containing 0.02% sodium azide.

Storage and Stability

Maintain refrigerated at 2-8°C in undiluted aliquots for up to 12 months.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enikő Sutus et al.
Scientific reports, 12(1), 2364-2364 (2022-02-13)
We have previously reported that RING1 and YY1 binding protein (RYBP) is important for central nervous system development in mice and that Rybp null mutant (Rybp-/-) mouse embryonic stem (ES) cells form more progenitors and less terminally differentiated neural cells
Paula Dobrinić et al.
Nature structural & molecular biology, 28(10), 811-824 (2021-10-06)
The Polycomb repressive system plays a fundamental role in controlling gene expression during mammalian development. To achieve this, Polycomb repressive complexes 1 and 2 (PRC1 and PRC2) bind target genes and use histone modification-dependent feedback mechanisms to form Polycomb chromatin
Izabella Bajusz et al.
Genes, 10(11) (2019-11-23)
Separation of germline cells from somatic lineages is one of the earliest decisions of embryogenesis. Genes expressed in germline cells include apoptotic and meiotic factors, which are not transcribed in the soma normally, but a number of testis-specific genes are
Rybp/DEDAF is required for early postimplantation and for central nervous system development.
Pirity, MK; Locker, J; Schreiber-Agus, N
Molecular and cellular biology null
Dynamic antagonism between key repressive pathways maintains the placental epigenome.
Weigert, et al.
Nature Cell Biology, 25, 579-591 (2023)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service