Skip to Content
Merck
All Photos(1)

Key Documents

AB1690

Sigma-Aldrich

Anti-Ubiquitin Antibody

serum, Chemicon®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

bovine, eukaryotes, human

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... UBAP2(55833)

Specificity

Ubiquitin (76 amino acid, 8.5 kDa). Shows staining of neurofibrillary tangles, neurites in senile plaques and a subset of neuronal processes. No reactivity seen to tau, neurofilament or microtubule-associated proteins.

Immunogen

Purified uboquitin covalently coupled to KLH using glutaraldehde.

Application

Anti-Ubiquitin Antibody detects level of Ubiquitin & has been published & validated for use in WB, IH, IH(P).
Immunohistochemistry: 1:1,000-1:5,000. Tissue fixed in 10% formalin, Bouin′s fixative for 1-2 days and embedded in paraffin, or unfixed frozen tissues may be used.

Immunoblotting: 1:1,000-1:5,000.

Optimal working dilutions must be determined by end user.
Research Category
Protein Trafficking
Research Sub Category
Ubiquitin & Ubiquitin Metabolism

Physical form

Neat rabbit antisera with 10 mM sodium azide.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jackalina M Van Kampen
Methods in molecular biology (Clifton, N.J.), 1806, 255-267 (2018-06-30)
Progranulin (PGRN) is a multifunctional protein that is widely expressed throughout the brain, where it has been shown to be a critical regulator of CNS inflammation (Ahmed et al., J Neuroinflammation 4:7, 2007; Yin et al., J Exp Med 207:117-128
Evaluation of selected binding domains for the analysis of ubiquitinated proteomes.
Nakayasu, ES; Ansong, C; Brown, JN; Yang, F; Lopez-Ferrer, D; Qian, WJ; Smith, RD; Adkins, JN
Journal of the American Society For Mass Spectrometry null
Peter Sutovsky et al.
Reproduction (Cambridge, England), 129(3), 269-282 (2005-03-08)
Major vault protein (MVP), also called lung resistance-related protein is a ribonucleoprotein comprising a major part (>70%) of the vault particle. The function of vault particle is not known, although it appears to be involved in multi-drug resistance and cellular
Nataša Bauer et al.
Frontiers in plant science, 10, 1469-1469 (2019-12-12)
MATH-BTB proteins are known to act as substrate-specific adaptors of CUL3-based E3 ligases in the ubiquitin proteasome pathway. Their BTB domain binds to CUL3 scaffold proteins and the less conserved MATH domain targets a highly diverse collection of substrate proteins
Ernesto S Nakayasu et al.
Journal of proteome research, 14(9), 4029-4038 (2015-07-07)
Ubiquitination is a key protein post-translational modification that regulates many important cellular pathways and whose levels are regulated by equilibrium between the activities of ubiquitin ligases and deubiquitinases. Here, we present a method to identify specific deubiquitinase substrates based on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service