Skip to Content
Merck
All Photos(3)

Key Documents

AB1582

Sigma-Aldrich

Anti-Choline Acetyltransferase Antibody

CHEMICON®, sheep polyclonal

Synonym(s):

ChAT

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

Product Name

Anti-Choline Acetyltransferase Antibody, serum, Chemicon®

biological source

sheep

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat, guinea pig, rabbit

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

Specificity

Choline Acetyltransferase (ChAT). Will stain cholinergic neurons in rat, guinea-pig and rabbit central and peripheral nervous systems.

Immunogen

A 22 amino acid peptide from porcine ChAT, coupled to KLH. The sequence is H-GLFSSYRLPGHTQDTLVAQKSS-NH2.

Application

Immunohistochemistry: 1:1,000-1:2,000

Immunoblotting

Optimal working dilutions must be determined by end user.

IMMUNOHISTOCHEMISTRY PROTOCOL FOR AB1582

STAINING OF GUINEA PIG ILEUM

1) Pin the ileum out flat and tight.

2) Fix overnight at 4°C in modified Zamboni′s (2% paraformaldehyde with 15% saturated picric acid in phosphate buffered saline).

3) Clear in DMSO, 100%, 3 times for 10 minutes each, rinse in PBS.

4) Dissect off ALL circular muscle.

5) Preincubate with 10% normal donkey serum.

6) Incubate in anti-ChAT (AB1582) diluted 1:1,000-1,2000 with 10% normal donkey serum over night at 4°C on a shaker.

7) Incubate with Donkey anti-Sheep IgG-Cy3 labeled secondary antibody (Catalog Number AP184C) at a dilution of 1:200 for 2 hours.

IMPORTANT NOTE: It is essential to remove ALL circular muscle.
Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors

Neuronal & Glial Markers
This Anti-Choline Acetyltransferase Antibody is validated for use in IH, WB for the detection of Choline Acetyltransferase.

Physical form

Sheep serum. Lyophilized, no preservatives. Reconstitute with 100 μL of sterile distilled water. Centrifuge to remove any residue.

Storage and Stability

Maintain at -20 to -70°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles. Glycerol (1:1) can be added for additional stability.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E E Hur et al.
Neuroscience, 164(4), 1721-1731 (2009-09-26)
The basal forebrain (BF) comprises morphologically and functionally heterogeneous cell populations, including cholinergic and non-cholinergic corticopetal neurons that are implicated in sleep-wake modulation, learning, memory and attention. Several studies suggest that glutamate may be among inputs affecting cholinergic corticopetal neurons
Guy R Sander et al.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 51(7), 969-972 (2003-06-18)
The Notch signaling pathway is a vitally important pathway in regulating brain development. To explore the involvement of the Notch pathway in neuronal cells of adult rat gut, we investigated the expression of Notch1 and Jagged2 by in situ hybridization
Cardiac Dysregulation and Myocardial Injury in a 6-Hydroxydopamine-Induced Rat Model of Sympathetic Denervation.
Jiang, YH; Jiang, P; Yang, JL; Ma, DF; Lin, HQ; Su, WG; Wang, Z; Li, X
Testing null
Geraldo E Vicentini et al.
PloS one, 11(9), e0162998-e0162998 (2016-09-17)
Gastrointestinal dysmotility frequently occurs in cancer cachexia and may result from damage to enteric innervation caused by oxidative stress, especially due to glutathione depletion. We assessed the effect of dietary supplementation with 20 g/kg l-glutamine (a glutathione precursor) on the
S D Motts et al.
Neuroscience, 154(1), 186-195 (2008-01-29)
We used an antibody to choline acetyltransferase (ChAT) to label cholinergic cells in guinea pig brainstem. ChAT-immunoreactive (IR) cells comprise several prominent groups, including the pedunculopontine tegmental nucleus, laterodorsal tegmental nucleus, and parabigeminal nucleus, as well as the cranial nerve

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service