Skip to Content
Merck
All Photos(1)

Key Documents

AB15708

Sigma-Aldrich

Anti-β-Tubulin III Antibody

CHEMICON®, rabbit polyclonal

Synonym(s):

Anti-CDCBM, Anti-CFEOM3, Anti-CFEOM3A, Anti-FEOM3, Anti-TUBB4, Anti-beta-4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

Product Name

Anti-β III tubulin Antibody, Chemicon®, from rabbit

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse

packaging

antibody small pack of 25 μg

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

mouse ... Tubb3(22152)

Related Categories

Specificity

Beta III Tubulin. By Western blot the antibody recognizes a band at ~50 kDa on mouse brain lysate. No reactivity with 3T3 cell lysate by Western blot.
It is expected that the antibody will react with most mammalian species.

Immunogen

Synthetic peptide from human beta III tubulin.

Application

Anti-β III tubulin Antibody is an antibody against β III tubulin for use in WB.
Research Category
Neuroscience
Research Sub Category
Neuronal & Glial Markers

Neurofilament & Neuron Metabolism
Western blot: 0.05-0.1 μg/mL on mouse brain lysate.

Optimal working dilutions must be determined by end user.

Quality

Tested

Physical form

Format: Purified
ImmunoAffinity Purified
Liquid in PBS with 0.1% sodium azide.

Storage and Stability

Maintain at 2-8°C in undiluted aliquots for up to 6 months after date of receipt.

Other Notes

Please refer to lot specific datasheet.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Neurite outgrowth induced by stimulation of angiotensin II AT2 receptors in SH-SY5Y neuroblastoma cells involves c-Src activation.
Blanco, et al.
Heliyon, 9, e15656-e15656 (2023)
Oscar Diaz-Horta et al.
Journal of molecular medicine (Berlin, Germany), 96(11), 1227-1238 (2018-10-04)
RIPOR2 (previously known as FAM65B) localizes to stereocilia of auditory hair cells and causes deafness when its function is disturbed by mutations. Here, we demonstrate that during the morphogenesis of the hair cell bundle, absence of Ripor2 affects the orientation
Victoria M Ho et al.
Molecular and cellular neurosciences, 61, 1-12 (2014-05-03)
AMPA-type glutamate receptors mediate fast, excitatory neurotransmission in the brain, and their concentrations at synapses are important determinants of synaptic strength. We investigated the post-transcriptional regulation of GluA2, the calcium-impermeable AMPA receptor subunit, by examining the subcellular distribution of its
Antoine G Almonte et al.
Journal of neurochemistry, 124(1), 109-122 (2012-11-02)
Protease-activated receptor-1 (PAR1) is an unusual G-protein coupled receptor (GPCR) that is activated through proteolytic cleavage by extracellular serine proteases. Although previous work has shown that inhibiting PAR1 activation is neuroprotective in models of ischemia, traumatic injury, and neurotoxicity, surprisingly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service