Skip to Content
Merck
All Photos(2)

Key Documents

AB153-I

Sigma-Aldrich

Anti-Fluorescent Gold Antibody

from rabbit

Synonym(s):

Fluorescent Marker

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity (predicted by homology)

all

technique(s)

immunohistochemistry: suitable (paraffin)

shipped in

wet ice

target post-translational modification

unmodified

General description

Fluorescent Gold (hydroxystilbamidine), better known as Fluorogold, has been a very popular fluorescent marker used to retrogradely label projection neurons. AB153 also reacts with aminostilbamidine quite nicely in frozen, 4% PFA fixed tissues.

Specificity

This antibody will react with any species sample treated with Fluorescent Gold.

Immunogen

KLH-conjugated Fluorescent Gold.

Application

Anti-Fluorescent Gold Antibody is a highly specific rabbit polyclonal antibody, that targets Fluoro-Gold & has been tested in IHC (Paraffin).
Immunohistochemistry Analysis: A representative lot from an independent laboratory detected Fluorescent Gold treated neurons in rat frontal cortex tissues (Chang, H. T., et al. (1990). J Neurosci Methods. 35(1):31-37.).

Quality

Evaluated by Immunohistochemistry in fixed frozen Fluorescent Gold treated rat cerebral cortex tissues.

Immunohistochemistry Analysis: A 1:10,000 dilution of this antibody detected Fluorescent Gold in fixed frozen Fluorescent Gold treated rat cerebral cortex tissues.

Physical form

Format: Purified

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Charles Raver et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 40(17), 3424-3442 (2020-03-29)
The parabrachial (PB) complex mediates both ascending nociceptive signaling and descending pain modulatory information in the affective/emotional pain pathway. We have recently reported that chronic pain is associated with amplified activity of PB neurons in a rat model of neuropathic
Sofia Meyer Zu Reckendorf et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 42(12), 2474-2491 (2022-02-13)
In postmitotic neurons, several tumor suppressor genes (TSGs), including p53, Rb, and PTEN, modulate the axon regeneration success after injury. Particularly, PTEN inhibition is a key driver of successful CNS axon regeneration after optic nerve or spinal cord injury. In
Anett J Nagy et al.
PLoS biology, 16(6), e2004712-e2004712 (2018-06-26)
Evolutionary development of vision has provided us with the capacity to detect moving objects. Concordant shifts of visual features suggest movements of the observer, whereas discordant changes are more likely to be indicating independently moving objects, such as predators or
Chang-Hui Chen et al.
Frontiers in neural circuits, 14, 605332-605332 (2020-12-17)
Area prostriata in primates has recently been found to play important roles in rapid detection and processing of peripheral visual, especially fast-moving visual information. The prostriata in rodents was not discovered until recently and its connectivity is largely unknown. As
Daniel Dautan et al.
Cell reports, 36(8), 109594-109594 (2021-08-26)
The mesencephalic locomotor region (MLR) serves as an interface between higher-order motor systems and lower motor neurons. The excitatory module of the MLR is composed of the pedunculopontine nucleus (PPN) and the cuneiform nucleus (CnF), and their activation has been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service