A1345
Antioxidant Supplement (1000×)
liquid, sterile-filtered, suitable for cell culture
Synonym(s):
Antioxidant Solution
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352205
NACRES:
NA.75
Recommended Products
sterility
sterile-filtered
form
liquid
technique(s)
cell culture | mammalian: suitable
impurities
endotoxin, tested
shipped in
ambient
storage temp.
2-8°C
General description
Antioxidant Supplement (1000×) is a proprietary medium supplement specially formulated to boost cell growth for enhanced viral and protein expression. It may be used in combination with Polyamine Supplement (P 8483).
One mL supplements 1 liter of medium.
One mL supplements 1 liter of medium.
Application
Antioxidant Supplement (1000×) has been used:
- as a component of the extender media for dilution of sperm samples during cryopreservation
- for long term culture of human-induced pluripotent stem (iPS) cells
- as a component of the immortalization media used for culturing, cloning and expansion of clones for making immortalized cell lines from embryonic mouse kidney
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Lan Luo et al.
Scientific reports, 4, 3779-3779 (2014-01-22)
Effects of antioxidants on the quality and genomic stability of induced pluripotent stem (iPS) cells were investigated with two human iPS cell lines (201B7 and 253G1). Cells used in this study were expanded from a single colony of each cell
E Cabrita et al.
Animal reproduction science, 125(1-4), 189-195 (2011-04-13)
During cryopreservation, dilution in the extender media reduces the seminal plasma constituents being cells more vulnerable to oxidative stress. Vitamins (C and E) and the amino acids taurine and hypotaurine are powerful antioxidants naturally present in seminal plasma. Whether their
Tullia Maraldi et al.
Rejuvenation research, 23(2), 93-106 (2019-08-02)
The efficient use of stem cells for transplantation is often limited by the relatively low number of stem cells collected. The ex vivo expansion of human stem cells for clinical use is a potentially valuable approach to increase stem cell
Michael R Duggan et al.
Cell death discovery, 7(1), 60-60 (2021-03-28)
Despite increasing numbers of aged individuals living with HIV, the mechanisms underlying HIV-associated neurological disorders (HANDs) remain elusive. As HIV-1 pathogenesis and aging are characterized by oxidative stress as well as altered protein quality control (PQC), reactive oxygen species (ROS)
Kendall Mitchell et al.
BMC neuroscience, 17(1), 80-80 (2016-12-03)
Bone morphogenetic protein-2 (BMP-2) is a pleiotropic, secreted molecule with diverse effects. The potent ability of BMP-2 to stimulate bone growth prompted its widespread clinical use for arthrodesis (spine fusion). However, elevated post-operative pain in patients treated with BMP-2 has
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service