Recommended Products
Quality Level
product line
NovaGel
Novabiochem®
form
beads
reaction suitability
reaction type: Fmoc solid-phase peptide synthesis
manufacturer/tradename
Novabiochem®
application(s)
peptide synthesis
functional group
amine
storage temp.
2-8°C
Related Categories
General description
This resin is prepared by derivatization of aminomethyl NovaGel with the modified Rink linker. NovaGel resins combine many of the excellent properties of PEG-PS-based resins with the high loading efficiency of conventional polystyrene-based supports. In a comparison with Rink amide resin, this resin was found to give superior results in the synthesis of peptides up to 17 amino acids in length [1]. The use of this support in the preparation of oligonucleotide-peptide conjugates has been described [2].
Associated Protocols and Technical Articles
Protocols for Loading of Peptide Synthesis Resins
Associated Protocols and Technical Articles
Protocols for Loading of Peptide Synthesis Resins
Linkage
Replaces: 01-64-0286
Analysis Note
Color (visual): white to yellow to beige
Appearance of substance (visual): beads
Loading (determined from the substitution of the Fmoc-Leu loaded resin): 0.50 - 0.70 mmol/g
Swelling Volume (in DMF): lot specific result
The PEG-PS resin is prepared from aminomethyl copoly(styrene-1 % DVB), 100 - 200 mesh
Appearance of substance (visual): beads
Loading (determined from the substitution of the Fmoc-Leu loaded resin): 0.50 - 0.70 mmol/g
Swelling Volume (in DMF): lot specific result
The PEG-PS resin is prepared from aminomethyl copoly(styrene-1 % DVB), 100 - 200 mesh
Legal Information
Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service