8.55001
Rink Amide resin (100-200 mesh)
Novabiochem®
Synonym(s):
4-(2′,4′-Dimethoxyphenyl-Fmoc-aminomethyl)-phenoxy resin
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Quality Level
product line
Novabiochem®
form
beads
reaction suitability
reaction type: Fmoc solid-phase peptide synthesis
manufacturer/tradename
Novabiochem®
application(s)
peptide synthesis
functional group
amine
storage temp.
2-30°C
General description
An excellent support for the Fmoc SPPS of peptide amides. This resin is more acid-sensitive than Rink Amide AM and Rink Amide MBHA resins. Cleavage with high concentrations of TFA can lead to the breakdown of the linker, with the concomitant formation of by-products that can not be removed by simple washes. These problems appear to be minimized through the use of low TFA concentrations or by the addition of trialkyl silanes to the cleavage mixture.
Associated Protocols and Technical Articles
Protocols for Loading of Peptide Synthesis Resins
Associated Protocols and Technical Articles
Protocols for Loading of Peptide Synthesis Resins
Linkage
Replaces: 01-64-0013
Analysis Note
Color (visual): white to pale yellow to beige
Appearance of substance (visual): beads
Loading (Photometric determination of the Fmoc-chromophore liberated upon treatment with DBU/DMF): 0.40 - 0.80 mmol/g
TNBS-Test: passes test
Swelling Volume (in DMF): lot specific result
Swelling Volume (in CH₂Cl₂): lot specific result
Total swelling volume acc. Houben Weyl (in CH2Cl2): ≥ 5.2 ml/g
Total swelling volume acc. Houben Weyl (in DMF): ≥ 5.2 ml/g
The polymer matrix is copoly (styrene-1 % DVB) 100 - 200 mesh.
Appearance of substance (visual): beads
Loading (Photometric determination of the Fmoc-chromophore liberated upon treatment with DBU/DMF): 0.40 - 0.80 mmol/g
TNBS-Test: passes test
Swelling Volume (in DMF): lot specific result
Swelling Volume (in CH₂Cl₂): lot specific result
Total swelling volume acc. Houben Weyl (in CH2Cl2): ≥ 5.2 ml/g
Total swelling volume acc. Houben Weyl (in DMF): ≥ 5.2 ml/g
The polymer matrix is copoly (styrene-1 % DVB) 100 - 200 mesh.
Legal Information
Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service