Skip to Content
Merck
All Photos(1)

Documents

8.53029

Sigma-Aldrich

Boc-Glu(OcHx)-OH

Novabiochem®

Synonym(s):

Boc-Glu(OcHx)-OH, N-α-t.-Boc-L-glutamic acid γ-cyclohexyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H27NO6
CAS Number:
Molecular Weight:
329.39
UNSPSC Code:
12352209
NACRES:
NA.22

Quality Level

product line

Novabiochem®

Assay

≥98% (TLC)

form

powder

reaction suitability

reaction type: Boc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

mp

54-57 °C

application(s)

peptide synthesis

functional group

carboxylic acid

storage temp.

2-30°C

InChI

1S/C16H27NO6/c1-16(2,3)23-15(21)17-12(14(19)20)9-10-13(18)22-11-7-5-4-6-8-11/h11-12H,4-10H2,1-3H3,(H,17,21)(H,19,20)/t12-/m0/s1

InChI key

FDNMLANBNJDIRG-LBPRGKRZSA-N

General description

Standard building block for introduction of glutamic acid in Boc SPPS. This side-chain protecting group for glutamic acid is used to minimize side reactions during acidic and basic treatments [1].

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Boc SPPS

Literature references

[1] J. P. Tam, et al. (1979) Tetrahedron Lett., 42, 4033.

Linkage

Replaces: 04-12-0121

Analysis Note

Colour (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Optical rotation α 25/D (c=1 in methanol): -9.0 - -7.0 °
Purity (TLC(0811)): ≥ 98 %
Purity (DC (0157)): ≥ 98 %
Solubility (1 mmole in 2 ml DMF): clearly soluble

To see the solvent systems used for TLC of Novabiochem® products please click here.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service