Skip to Content
Merck
All Photos(1)

Key Documents

8.41705

Sigma-Aldrich

3-Mercapto-1,2-propanediol

for synthesis

Synonym(s):

3-Mercapto-1,2-propanediol, 1-Thioglycerol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H8O2S
CAS Number:
Molecular Weight:
108.16
MDL number:
UNSPSC Code:
12352104
EC Index Number:
202-495-0
NACRES:
NA.22

vapor pressure

<1 hPa ( 20 °C)

Quality Level

form

liquid

pH

4.79 (20 °C, 10 g/L in H2O)

mp

<-20 °C

transition temp

flash point 99 °C

solubility

slightly soluble >1000 g/L

density

1.247 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

SCC(O)CO

InChI

1S/C3H8O2S/c4-1-3(5)2-6/h3-6H,1-2H2

InChI key

PJUIMOJAAPLTRJ-UHFFFAOYSA-N

Application

  • Polyhydroxyurethanes and Semi-IPN Hydrogels: 3-Mercapto-1,2-propanediol is utilized in the simultaneous formation of polyhydroxyurethanes and multicomponent semi-interpenetrating polymer networks (semi-IPN) hydrogels, enhancing material properties for diverse biomedical applications (Carbajo-Gordillo et al., 2024).
  • Nanoparticle Functionalization: This compound facilitates the direct binding and characterization of enzymes onto iron oxide nanoparticles, showcasing its role in nanotechnology for biotechnological applications (Sidhu et al., 2024).
  • Shape Memory Polyurethanes: Used in the synthesis of multifunctional polyols via a facile thiol-ene click reaction method (Lee and Kim, 2023).
  • Low-Temperature Nanophosphor Synthesis: It is instrumental in the novel low-temperature synthesis of europium-doped yttrium oxide nanophosphors, utilized in near-UV light-emitting diodes, highlighting its critical role in advanced material manufacturing (Patwardhan et al., 2023).
  • Phosphorylcholine monomer: 3-Mercapto-1,2-propanediol is used in the synthesis of functional phosphorylcholine monomer of 2-((3-((2,3-dihydroxypropyl)thio)-2-methylpropanoyl)oxy)ethyl phosphorylcholine (PC-diol)via a click reaction (Hao et al., 2023).

Analysis Note

Assay (iodometric): ≥ 98.0 %
Identity (IR): passes test

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Skin Irrit. 2 - Skin Sens. 1B

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

210.2 °F - closed cup

Flash Point(C)

99 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service