Skip to Content
Merck
All Photos(1)

Key Documents

8.41355

Sigma-Aldrich

Isophthalonitrile

for synthesis

Synonym(s):

Isophthalonitrile, Benzene-1,3-dicarbonitrile, 1,3-Dicyanobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H4N2
CAS Number:
Molecular Weight:
128.13
MDL number:
UNSPSC Code:
12352117
EC Index Number:
210-933-7
NACRES:
NA.22

vapor pressure

0.014 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

form

powder

potency

>2000 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)

bp

288 °C/1013 hPa

mp

162-163 °C

density

1.3 g/cm3 at 20 °C

bulk density

500 kg/m3

storage temp.

2-30°C

SMILES string

N#Cc1cc(ccc1)C#N

InChI

1S/C8H4N2/c9-5-7-2-1-3-8(4-7)6-10/h1-4H

InChI key

LAQPNDIUHRHNCV-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 159 °C
Melting range (upper value): ≤ 162 °C
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service