Skip to Content
Merck
All Photos(2)

Key Documents

145858

Sigma-Aldrich

1,3-Dicyanobenzene

98%

Synonym(s):

Isophthalodinitrile, Isophthalonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(CN)2
CAS Number:
Molecular Weight:
128.13
Beilstein:
1072209
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

mp

163-165 °C (lit.)

SMILES string

N#Cc1cccc(c1)C#N

InChI

1S/C8H4N2/c9-5-7-2-1-3-8(4-7)6-10/h1-4H

InChI key

LAQPNDIUHRHNCV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1,3-Dicyanobenzene was used in the synthesis of oxazolines and 1,3-bis-(4,4-dimethyl-2-oxazolinyl)benzene.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luo Mei et al.
Journal of combinatorial chemistry, 11(2), 220-227 (2009-01-14)
A class of modular oxazolines and their derivatives 1-5 were synthesized with moderate to excellent yields using a simple one-pot method; 4 x 3 bis-oxazolines were obtained, as expected, from each of the three reactions of 1,4-dicyanobenzene, 1,3-dicyanobenzene, and 1,2-dicyanobenzene
Glenn R Masson et al.
The Biochemical journal, 474(11), 1867-1877 (2017-04-07)
Until recently, one of the major limitations of hydrogen/deuterium exchange mass spectrometry (HDX-MS) was the peptide-level resolution afforded by proteolytic digestion. This limitation can be selectively overcome through the use of electron-transfer dissociation to fragment peptides in a manner that
A SIMPLE LARGE SCALE SYNTHESIS OF 1, 3-BIS-(4, 4-DIMETHYL-2-OXAZOLINYL) BENZENE 1*.
Button KM, et al.
Synthetic Communications, 32(3), 363-368 (2002)
Marcin Konowalczyk et al.
Physical chemistry chemical physics : PCCP, 23(2), 1273-1284 (2020-12-24)
Modulated magnetically altered reaction yield (ModMARY) spectroscopy is a derivative variant of fluorescence detected magnetic field effect measurement, where the applied magnetic field has both a constant and a modulated component. As in many derivative spectroscopy techniques, the signal to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service