Skip to Content
Merck
All Photos(1)

Key Documents

8.20092

Sigma-Aldrich

4-Hydroxyanilinium chloride

for synthesis

Synonym(s):

4-Hydroxyanilinium chloride, 4-Aminophenol hydrochloride, 4-Hydroxyaniline hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H7NO · xHCl
CAS Number:
Molecular Weight:
109.13 (free base basis)
MDL number:
UNSPSC Code:
12352107
EC Index Number:
200-122-6
NACRES:
NA.22

Quality Level

mp

306 °C (decomposition)

storage temp.

2-30°C

SMILES string

[Cl-].[N+H3]c1ccc(cc1)O

InChI

1S/C6H7NO.ClH/c7-5-1-3-6(8)4-2-5;/h1-4,8H,7H2;1H

InChI key

RVGOBWDGAVAVPJ-UHFFFAOYSA-N

Analysis Note

Assay (argentometric): ≥ 99.0 %
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Carc. 2 - Eye Dam. 1 - Muta. 2 - Skin Sens. 1 - STOT RE 1

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service