Skip to Content
Merck
All Photos(1)

Key Documents

8.19040

Sigma-Aldrich

Ethylenediaminetetraacetic acid dipotassium salt dihydrate

for synthesis

Synonym(s):

Ethylenediaminetetraacetic acid dipotassium salt dihydrate, Ethylenedinitrilotetraacetic acid dipotassium salt, Titriplex® Potassium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14K2N2O8 · 2H2O
CAS Number:
Molecular Weight:
404.45
MDL number:
UNSPSC Code:
12352107
EC Index Number:
217-895-0
NACRES:
NA.22

Quality Level

form

solid

potency

>2000 mg/kg LD50, oral (Rat)

pH

4-5 (20 °C, 50 g/L in H2O)

mp

272 °C

bulk density

310 kg/m3

storage temp.

2-30°C

SMILES string

[K+].[K+].N(CCN(CC(=O)O)CC(=O)O)(CC(=O)[O-])CC(=O)[O-].O.O

InChI

1S/C10H16N2O8.2K.2H2O/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20;;;;/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20);;;2*1H2/q;2*+1;;/p-2

InChI key

JFROQFOCFOKDKU-UHFFFAOYSA-L

Related Categories

Analysis Note

Assay (complexometric; calculated on dried substance): 99.0 - 101.5 %
Identity (IR): passes test

Legal Information

Titriplex is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Inhalation

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service