Skip to Content
Merck
All Photos(5)

Key Documents

795410

Sigma-Aldrich

Sodium phosphate dibasic

anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%

Synonym(s):

sec-Sodium phosphate, Disodium hydrogen phosphate, Disodium phosphate, Sodium hydrogenphosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2HPO4
CAS Number:
Molecular Weight:
141.96
EC Number:
MDL number:
UNSPSC Code:
12161705
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥99%
grade:
ACS reagent
anhydrous
form:
powder

grade

ACS reagent
anhydrous

Quality Level

Agency

suitable for EPA 533
suitable for SM 5210

product line

Redi-Dri

Assay

≥99%

form

powder

quality

free-flowing

impurities

≤0.002% Chloride (Cl)
≤0.01% Insoluble matter

loss

≤0.2% loss on drying, 105°C

pH

8.7-9.3 (25 °C, 5% in solution)

pKa (25 °C)

(1) 2.15, (2) 6.82, (3) 12.38 (phosphoric acid)

anion traces

sulfate (SO42-): ≤0.005%

cation traces

Fe: ≤0.002%
heavy metals: ≤0.001% (by ICP-OES)

SMILES string

[Na+].[Na+].[H]OP([O-])([O-])=O

InChI

1S/2Na.H3O4P/c;;1-5(2,3)4/h;;(H3,1,2,3,4)/q2*+1;/p-2

InChI key

BNIILDVGGAEEIG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Redi-Dri technology eliminates clumping and the need for complicated procedures to keep your salts free-flowing. Keeping your salts dry helps improve your results by giving accurate measurements and easier-to-handle salts and buffers. Rather than drying your salts in an oven or desiccator or breaking them apart manually, Redi-Dri ensures that your salts will remain easy-to-use for months without worrying about anti-caking agents. Even after repeated openings you can be assured that your results are always accurate. Redi-Dri saves you time, energy and money by keeping your salts usable for longer!
Sodium phosphate dibasic is an inorganic salt with the chemical formula Na2HPO4. Sodium phosphate dibasic is highly soluble in water but insoluble in ethanol. It is used as a sequestering agent, buffering agent, pH regulator and saline laxative.

Application

Sodium phosphate dibasic can be used as a buffer in the reverse phase HPLC method.

Features and Benefits

  • Increased efficiency – Eliminate time and effort of loosening hardened powders
  • Safety – Avoid risks of using tools of force to break up clumps
  • Economy – Faster preparation and solvation increase productivity and reduce costs
  • Assured quality – Excellent, expert-tested quality with no anti-caking agents
  • Flexible volumes – Available from research amounts to scale-up quantities.

  • Increased efficiency – Eliminate time and effort of loosening hardened powders
  • Safety – Avoid risks of using tools of force to break up clumps
  • Economy – Faster preparation and solvation increase productivity and reduce costs
  • Assured quality – Excellent, expert-tested quality with no anti-caking agents
  • Flexible volumes – Available from research amounts to scale-up quantities

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rachana Srivastava et al.
International journal of biological macromolecules, 145, 252-261 (2019-12-25)
To evaluate the role of micelles for protein-surfactant interaction, we have studied the binding modes of serum albumin proteins (human (HSA) and rabbit (RSA)) with anionic-surfactant, sodium dodecyl sulfate (SDS) by using UV-visible, fluorescence, circular dichroism, fluorescence lifetime, atomic force

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service