Skip to Content
Merck
All Photos(1)

Key Documents

81066N

Sigma-Aldrich

Probumin® Bovine Serum Albumin Reagent Grade

≥98%, USA origin, 0.0-7.0 mg/dL Urea Nitrogen, enzyme immunoassay: suitable, Serology: suitable, Protein standard

Synonym(s):

Albumin, BSA, Bovine Serum, Bovine Serum Albumin, Fraction V

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41141801
NACRES:
NA.28

Product Name

Probumin® Bovine Serum Albumin Reagent Grade, pack of 10 g

biological source

bovine serum

Quality Level

grade

reagent grade

Assay

≥98% (purity)

form

lyophilized powder

shelf life

48 mo.

purified by

heat shock fractionation

packaging

pack of 10 g

manufacturer/tradename

Probumin®

origin

USA origin

free fatty acid content content

0.0-1.0 mg (per gram)

Total lipids content

0.0-3.5 mg (per gram)

technique(s)

enzyme immunoassay: suitable
microbiological culture: suitable
protein quantification: suitable

dilution

( Serology)

impurities

0.0-0.5 mg/g L-Lactate
0.0-4.0 mg/dL Uric Acid
0.0-7.0 mg/dL Urea Nitrogen

loss

≤5%

pH range

6.8-7.2

anion traces

chloride (Cl-): 0.0-3.0 mg/g

cation traces

Fe: 0.0-5.0 μg/g
Na: 0.0-10.0 mg/g
heavy metals: 0.0-20.0 μg/g (Cu, Fe, Pb, Zn)

foreign activity

Tested for IgG

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Manufactured by a proprietary heat-shock fractionation process, using caprylic acid as an albumin stabilizer. This BSA powder is solvent treated and extensively diafiltered. This bovine albumin product is low in sodium and purity is >=98%; typical purity is 100%.

Legal Information

Probumin is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Probumin® BSA grades are distinguished by differences in production. The use of terms such as ′Diagnostic Grade′ or ′Vaccine Grade′ in product names solely indicates that said products are commonly used by customers in those applications. We have not qualified every single product in every conceivable application. Users should qualify Probumin® BSA grades using the specific reagents, conditions, and constraints of their assay.
For further manufacturing use only. Not for human or animal consumption.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service