8.00841
Methyl acrylate
(stabilised with hydroquinone monomethyl ether) for synthesis
Synonym(s):
Methyl acrylate, Methyl propenoate, Acrylic acid methyl ester
About This Item
Recommended Products
grade
for synthesis
Quality Level
vapor density
3 (vs air)
vapor pressure
67.5 mmHg ( 20 °C)
Assay
≥99% (GC)
form
liquid
autoignition temp.
874 °F
expl. lim.
14.5 %
dilution
(for synthesis)
refractive index
n20/D 1.402 (lit.)
pH
7 (20 °C)
kinematic viscosity
10 cSt(23 °C)
bp
80 °C (lit.)
mp
−75 °C (lit.)
transition temp
flash point -2.7 °C
solubility
60 g/L
density
0.956 g/mL at 25 °C (lit.)
storage temp.
2-30°C
SMILES string
COC(=O)C=C
InChI
1S/C4H6O2/c1-3-4(5)6-2/h3H,1H2,2H3
InChI key
BAPJBEWLBFYGME-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Analysis Note
Density (d 20 °C/ 4 °C): 0,954 - 0,958
Identity (IR): conforms
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
27.0 °F - closed cup
Flash Point(C)
-2.8 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service