Skip to Content
Merck
All Photos(1)

Key Documents

658401

Sigma-Aldrich

AG 490

A cell-permeable, reversible, substrate competitive, and potent inhibitor of epidermal growth factor receptor kinase autophosphorylation (IC₅₀ = 100 nM).

Synonym(s):

AG 490, α-Cyano-(3,4-dihydroxy)-N-benzylcinnamide, Tyrphostin B42, ( E)- N-benzyl-2-cyano-3-(3,4-dihydroxyphenyl)acrylamide, JAK1 Inhibitor II, JAK2 Inhibitor VI, JAK3 Inhibitor XI, α-Cyano-(3,4-dihydroxy)-N-benzylcinnamide, Tyrphostin B42, (E)-N-benzyl-2-cyano-3-(3,4-dihydroxyphenyl)acrylamide, JAK1 Inhibitor II, JAK2 Inhibitor VI, JAK3 Inhibitor XI

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H14N2O3
CAS Number:
Molecular Weight:
294.30
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 100 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

N(Cc2ccccc2)C(=O)\C(=C\c1cc(c(cc1)O)O)\C#N

InChI

1S/C17H14N2O3/c18-10-14(8-13-6-7-15(20)16(21)9-13)17(22)19-11-12-4-2-1-3-5-12/h1-9,20-21H,11H2,(H,19,22)/b14-8+

InChI key

TUCIOBMMDDOEMM-RIYZIHGNSA-N

General description

A cell-permeable, reversible, substrate competitive, and potent inhibitor of epidermal growth factor receptor kinase autophosphorylation (IC50 = 100 nM). Inhibition of JAK2 by AG 490 selectively blocks leukemic cell growth in vitro and in vivo by inducing programmed cell death, with no harmful effect on normal hematopoiesis. Inhibits the constitutive activation of STAT-3 DNA binding and IL-2-induced growth of MF tumor cells. AG 490 has also been shown to inhibit the autokinase activity of JAK3. A 100 mM (5 mg/170 µl) solution of AG490 (Cat. No. 658411) in DMSO is also available.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
epidermal growth factor receptor kinase autophosphorylation
Product does not compete with ATP.
Reversible: yes
Target IC50: 100 nM against epidermal growth factor receptor kinase autophosphorylation

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-70°C). Stock solutions are stable for up to 6 months at -70°C.

Other Notes

Jaleel, M., et al. 2004. Biochemistry43, 8247.
Eriksen, K.W., et al. 2001. Leukemia15, 787.
Kirken, R.A., et al. 1999. Leukoc. Biol.65, 891.
Nielsen, M., et al. 1997. Proc. Natl. Acad. Sci. USA 94, 6764.
Meydan, N., et al. 1996. Nature 379, 645.
Gazit, A., et al. 1991. J. Med. Chem.34, 1896.
Levitzki, A. 1990. Biochem. Pharmacol.40, 913.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fang Liu et al.
Neuropsychiatric disease and treatment, 17, 2489-2498 (2021-08-07)
Clinically, electroacupuncture (EA) improves cerebral ischemic injury, but its mechanism remains unknown. The aim of this study was to confirm the protective effects of EA on focal cerebral ischemia (FCI)-induced injury and the possible mechanism. Sprague-Dawley (SD) rats served as
Yuki Maeda et al.
Cell reports, 42(5), 112520-112520 (2023-05-18)
Oscillatory Hes1 expression activates cell proliferation, while high and sustained Hes1 expression induces quiescence, but the mechanism by which Hes1 differentially controls cell proliferation depending on its expression dynamics is unclear. Here, we show that oscillatory Hes1 expression down-regulates the
Rosemary Li et al.
Diabetes, 69(7), 1463-1475 (2020-04-26)
Diabetes occurs due to a loss of functional β-cells, resulting from β-cell death and dysfunction. Lactogens protect rodent and human β-cells in vitro and in vivo against triggers of β-cell cytotoxicity relevant to diabetes, many of which converge onto a
Beatriz Rendon-Mitchell et al.
Journal of immunology (Baltimore, Md. : 1950), 170(7), 3890-3897 (2003-03-21)
We recently discovered that a ubiquitous protein, high mobility group box 1 protein (HMGB1), is released by activated macrophages, and functions as a late mediator of lethal systemic inflammation. To elucidate mechanisms underlying the regulation of HMGB1 release, we examined
Sonia Nasi et al.
Annals of the rheumatic diseases, 75(7), 1372-1379 (2015-08-09)
Basic calcium phosphate (BCP) crystal and interleukin 6 (IL-6) have been implicated in osteoarthritis (OA). We hypothesise that these two factors may be linked in a reciprocal amplification loop which leads to OA. Primary murine chondrocytes and human cartilage explants

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service