Skip to Content
Merck
All Photos(1)

Key Documents

572630

Sigma-Aldrich

SU-5402

≥95% (HPLC), lyophilized solid, FGFR1 inhibitor, Calbiochem

Synonym(s):

SU5402, 3-[3-(2-Carboxyethyl)-4-methylpyrrol-2-methylidenyl]-2-indolinone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H16N2O3
CAS Number:
Molecular Weight:
296.32
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

SU5402, SU5402, CAS 215543-92-3, is a cell-permeable, reversible, and ATP-competitive inhibitor of the tyrosine kinase activity of FGFR1 (IC₅₀ = 10-20 µM in the presence of 1 mM ATP).

Quality Level

Assay

≥95% (HPLC)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

solubility

DMSO: 25 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[nH]1c(c(c(c1)C)CCC(=O)O)\C=C2\c3c(cccc3)NC\2=O

InChI

1S/C17H16N2O3/c1-10-9-18-15(11(10)6-7-16(20)21)8-13-12-4-2-3-5-14(12)19-17(13)22/h2-5,8-9,18H,6-7H2,1H3,(H,19,22)(H,20,21)/b13-8-

InChI key

JNDVEAXZWJIOKB-JYRVWZFOSA-N

General description

A cell-permeable, reversible, and ATP-competitive inhibitor of the tyrosine kinase activity of fibroblast growth factor receptor 1 (FGFR1; IC50 = 10-20 µM in the presence of 1 mM ATP). Also inhibits aFGF-induced tyrosine phosphorylation of ERK1 and ERK2 (IC50 = 10-20 µM). In contrast to SU4984 (Cat. No. 572625), SU5402 is only a weak inhibitor of tyrosine phosphorylation of the PDGF receptor and does not inhibit phosphorylation of the insulin receptor. Does not inhibit the kinase activity of the EGF receptor. A 10 mM (500 µg/169 µl) solution of SU5402 (Cat. No. 572631) in DMSO is also available.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Fibroblast growth factor receptor 1 (FGFR1)
Product competes with ATP.
Reversible: yes
Target IC50: 10-20 &10-20 µM for fibroblast growth factor receptor 1 (FGFR1) in the presence of 1 mM ATP;M against FGFR1 in the presence of 1 mM ATP; 10-20 µM against inhibits aFGF-induced tyrosine phosphorylation of ERK1 and ERK2

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Mohammadi, M., et al. 1997. Science 276, 955.

Legal Information

Not available for sale in the United States.
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shuyan Yang et al.
Heliyon, 9(11), e21915-e21915 (2023-11-30)
Nkx2.3, a transcription factor, plays important roles in various developmental processes. However, the mechanisms underlying nkx2.3's regulation of pouch and pharyngeal arch development in zebrafish remain unclear. In this study, we demonstrated that knockdown or knockout of nkx2.3 resulted in
H J Standley et al.
Development (Cambridge, England), 128(8), 1347-1357 (2001-03-23)
The community effect is an interaction among a group of many nearby precursor cells, necessary for them to maintain tissue-specific gene expression and differentiate co-ordinately. During Xenopus myogenesis, the muscle precursor cells must be in group contact throughout gastrulation in
Danilo Ranieri et al.
Biomedicines, 10(7) (2022-07-28)
Pancreatic ductal adenocarcinoma (PDAC) is a lethal malignancy whose main characterizations are Kirsten Rat Sarcoma-activating mutations (KRAS) and a highly aggressive phenotype. Based on our recent findings demonstrating that the highly aberrant expression of the mesenchymal isoform of Fibroblast Growth
Claire Leist Hinsch et al.
Cancers, 15(24) (2023-12-23)
How human FGFR1 localizes to the PM is unknown. Currently, it is assumed that newly synthesized FGFR1 is continuously delivered to the PM. However, evidence indicates that FGFR1 is mostly sequestered in intracellular post-Golgi vesicles (PGVs) under normal conditions. In
Julia Leschik et al.
Nature protocols, 3(9), 1381-1387 (2008-09-06)
Primate nonhuman and human embryonic stem (ES) cells provide a powerful model of early cardiogenesis. Furthermore, engineering of cardiac progenitors or cardiomyocytes from ES cells offers a tool for drug screening in toxicology or to search for molecules to improve

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service