Skip to Content
Merck
All Photos(2)

Key Documents

553030

Sigma-Aldrich

FPS-ZM1

≥98% (HPLC), semisolid,viscous liquid, RAGE antagonist, Calbiochem®

Synonym(s):

RAGE Antagonist, FPS-ZM1, Receptor for AGE Antagonist, FPS-ZM1, N-Benzyl-4-chloro-N-cyclohexylbenzamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H22ClNO
CAS Number:
Molecular Weight:
327.85
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Product Name

RAGE Antagonist, FPS-ZM1, RAGE Antagonist, FPS-ZM1, is a blood-brain-barrier permeant blocker of RAGE V domain-mediated ligand binding (Ki = 25, 148, & 230 nM, respectively, against Aβ40, HMGB1 & S100B, binding to sRAGE).

Quality Level

Assay

≥98% (HPLC)

form

semisolid
viscous liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white to off-white

solubility

DMSO: 100 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

O=C(C1=CC=C(Cl)C=C1)N(CC2=CC=CC=C2)C3CCCCC3

InChI

1S/C20H22ClNO/c21-18-13-11-17(12-14-18)20(23)22(19-9-5-2-6-10-19)15-16-7-3-1-4-8-16/h1,3-4,7-8,11-14,19H,2,5-6,9-10,15H2

InChI key

XDFKWGIBQMHSOH-UHFFFAOYSA-N

General description

A blood-brain-barrier-permeant, non-toxic, tertiary amide compound that acts as a high affinity, potent, multimodal blocker of RAGE (Receptor for Advanced Glycation End products) V domain-mediated ligand binding (Ki = 25, 148, and 230 nM, respectively, against Aβ40, HMGB1, and S100B, binding to sRAGE). Blocks RAGE-mediated influx of Aβ40 and Aβ42 into the brain. Also shown to suppress Aβ-RAGE induced NF-κB activation and NF-κB-dependent transcription of β-secretase. Daily treatment of APPsw/0 murine AD model (1 mg/kg/d via i.p.) is reported to greatly reduce Thioflavin S-positive amyloid plaques in cortex and hippocampus (by 70 to 80%) and restore congnitive performance to the level of non-AD mice.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
RAGE
Reversible: yes
Target Ki: 25, 148, and 230 nM, respectively, against A&beta

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Deane, R., et al. 2012. J. Clin. Invest.122, 1377.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Muskan Rajkumar Thakur et al.
Journal of biochemical and molecular toxicology, 38(3), e23664-e23664 (2024-02-19)
The present work elucidates the role of colchicine (COL) on albumin glycation and cellular oxidative stress in diabetic cardiomyopathy (DCM). Human serum albumin (HSA) was glycated with methylglyoxal in the presence of COL (2.5, 3.75, and 5 µM), whereas positive and negative
Chao Song et al.
Brain sciences, 12(5) (2022-05-29)
(1) Background: Chemotherapy-induced cognitive impairment (CICI) is often observed in cancer patients and impairs their life quality. Grape-seed-orientated procyanidin has been shown to have anti-inflammatory and neuroprotective effects, yet its effects in preventing CICI have not been investigated. (2) Method:
Han-Kyul Nam et al.
Biological & pharmaceutical bulletin, 44(11), 1697-1706 (2021-11-02)
Advanced glycation end products (AGEs) are the products formed through a non-enzymatic reaction of reducing sugars with proteins or lipids. There is a potential for toxicity in the case of AGEs produced through glycation with dicarbonyl compounds including methylglyoxal, glyoxal
Vijayasree V Giridharan et al.
Neurotherapeutics : the journal of the American Society for Experimental NeuroTherapeutics, 18(1), 640-653 (2020-09-05)
Pneumococcal meningitis is a life-threatening infection of the central nervous system (CNS), and half of the survivors of meningitis suffer from neurological sequelae. We hypothesized that pneumococcal meningitis causes CNS inflammation via the disruption of the blood-brain barrier (BBB) and
Yidi Wu et al.
American journal of physiology. Lung cellular and molecular physiology, 319(1), L185-L195 (2020-05-21)
S100A4 is a low-molecular-mass (12 kDa) EF-hand Ca2+-binding S100 protein that is expressed in a broad range of normal tissue and cell types. S100A4 can be secreted from some cells to act in an autocrine or paracrine fashion on target

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service