Skip to Content
Merck
All Photos(1)

Key Documents

5.30409

Sigma-Aldrich

TLR8 Agonist, cpd14b

Synonym(s):

TLR8 Agonist, cpd14b, Toll-Like Receptor 8 Agonist

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H18N2
CAS Number:
Molecular Weight:
214.31
UNSPSC Code:
51111800
NACRES:
NA.77

Assay

≥98% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

General description

A pentyl-quinoline derived compound that acts as a pure, potent agonist of Toll-like receptor 8 agonist (TLR8, EC50 = 200 nM in a human TLR8-specifc reporter gene assay). Does not affect TLR2, 3, 4, 5, 7, 9, 10, Nod1 and Nod2, and is devoid of any TLR7 activity. Shown to induce activities of several cytokines and chemokines, including IL-1a, IL-1b, IL-6, IL-8, IL-10, IL12, IL-18, IFN-g, CCL20, RANTES, GRO, GM-CSF, and Eotaxin (~ 10 mg/ml).
A pentyl-quinoline derived compound that acts as a pure, potent agonist of Toll-like receptor 8 agonist (TLR8, EC50 = 200 nM in a human TLR8-specifc reporter gene assay). Does not affect TLR2, 3, 4, 5, 7, 9, 10, Nod1 and Nod2, and is devoid of any TLR7 activity. Shown to induce activities of several cytokines and chemokines, including IL-1a, IL-1b, IL-6, IL-8, IL-10, IL12, IL-18, IFN-g, CCL20, RANTES, GRO, GM-CSF, and Eotaxin (~ 10 mg/ml).

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
TLR8
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Other Notes

Kokatla, H. P., et al. 2014, ChemMedChem9, 719.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service