Skip to Content
Merck
All Photos(1)

Key Documents

SML2566

Sigma-Aldrich

CL097

≥98% (HPLC)

Synonym(s):

2-(Ethoxymethyl)-3H-imidazo[4,5-c]quinolin-4-amine, CL 097, CL-097

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H14N4O
CAS Number:
Molecular Weight:
242.28
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to very dark brown

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

2-8°C

Biochem/physiol Actions

CL097 is an imidazoquinoline (IMDQ) derivative (IQD) with dual toll-like receptor 7 & 8 (TLR7 & TLR8; TLR7/8) agonist activity. CL097 is commonly employed in the concentration range from 0.5 μg/mL to 45 μg/mL for stimulating cell surface TLR7/8 in cultures and is reported to be a stronger TLR7 ligand than CL075.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jueqi Chen et al.
Nature, 564(7734), 71-76 (2018-11-30)
The NLRP3 inflammasome, which has been linked to human inflammatory diseases, is activated by diverse stimuli. How these stimuli activate NLRP3 is unknown. Here we show that different NLRP3 stimuli lead to disassembly of the trans-Golgi network (TGN). NLRP3 is
Zhikuan Zhang et al.
Cell reports, 25(12), 3371-3381 (2018-12-20)
Toll-like receptor 7 (TLR7) is an innate immune receptor for single-stranded RNA (ssRNA) and has important roles in infectious diseases. We previously reported that TLR7 shows synergistic activation in response to two ligands, guanosine and ssRNA. However, the specific ssRNA
Masaaki Okamoto et al.
The Journal of biological chemistry, 293(48), 18585-18600 (2018-10-05)
The innate immune system is important for the efficacy of vaccines, but excessive innate immune responses can cause adverse reactions after vaccination. Extracellular vesicles (EVs) are enriched in the blood and can deliver functional RNAs, such as microRNAs (miRNAs), to
Susanne Maria Ziegler et al.
Journal of reproductive immunology, 128, 30-37 (2018-06-11)
During pregnancy the maternal immune system has to develop tolerance towards the developing fetus. These changes in maternal immunity can result in increased severity of certain infections, but also in amelioration of autoimmune diseases. Pregnancy-related hormones have been suggested to
A León-Flores et al.
Immunology letters, 203, 70-79 (2018-09-22)
Recent evidence has revealed that PD-L1 is expressed in two functional forms, namely, a membrane-bound form (mPD-L1) and a soluble form (sPD-L1). The identification of the soluble form of PD-L1 represents the discovery of a new potential mechanism for the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service