Skip to Content
Merck
All Photos(1)

Key Documents

5.08408

Sigma-Aldrich

Batimastat

InSolution, ≥97%, 25 mM in DMSO

Synonym(s):

InSolution Batimastat, (4-N-Hydroxyamino)-2 R-isobutyl-3 S-(thienylthiomethyl)succinyl)-L-phenylalanine-N-methylamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H31N3O4S2
CAS Number:
Molecular Weight:
477.64
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥97% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles
desiccated (hygroscopic)
protect from light

storage temp.

−70°C

SMILES string

[s]1c(ccc1)SC[C@@H]([C@@H](CC(C)C)C(=O)N[C@@H](Cc2ccccc2)C(=O)NC)C(=O)NO

InChI

1S/C23H31N3O4S2/c1-15(2)12-17(18(22(28)26-30)14-32-20-10-7-11-31-20)21(27)25-19(23(29)24-3)13-16-8-5-4-6-9-16/h4-11,15,17-19,30H,12-14H2,1-3H3,(H,24,29)(H,25,27)(H,26,28)/t17-,18+,19+/m1/s1

InChI key

XFILPEOLDIKJHX-QYZOEREBSA-N

Biochem/physiol Actions

Cell permeable: yes
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Physical form

A 25 mM (2 mg/168 µL) sterile-filtered solution of Batimastat (Cat. No. 196440) in DMSO.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

188.6 °F - (refers to pure substance)

Flash Point(C)

87 °C - (refers to pure substance)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service