Skip to Content
Merck
All Photos(1)

Documents

444225

Sigma-Aldrich

MMP-3 Inhibitor II

The MMP-3 Inhibitor II, also referenced under CAS 161314-17-6, controls the biological activity of MMP-3. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Synonym(s):

MMP-3 Inhibitor II, N-Isobutyl-N-(4-methoxyphenylsulfonyl)-glycylhydroxamic Acid, NNGH

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H20N2O5S
CAS Number:
Molecular Weight:
316.37
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (TLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

DMSO: 30 mg/mL
ethanol: 8 mg/mL

shipped in

ambient

storage temp.

10-30°C

General description

A cell-permeable, potent inhibitor of human MMP-3 (stromelysin; Ki = 130 nM).
A potent and cell-permeable inhibitor of human MMP-3 (Ki = 130 nM).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
human MMP-3
Product does not compete with ATP.
Reversible: no
Target Ki: 130 nM against human MMP-3

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Kato, N., et al. 2002. Genome Informatics13, 272.
MacPherson, L.J., et al. 1997. J. Med. Chem. 40, 2525.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Angelika K Lemke et al.
Cell and tissue research, 340(1), 179-188 (2010-03-11)
Pro-inflammatory cytokines induce meniscal matrix degradation and inhibition of endogenous repair mechanisms, but the pathogenic mechanisms behind this are mostly unknown. Therefore, we investigated details of interleukin-1 (IL-1alpha)-induced aggrecan turnover in mature meniscal tissue explants. Fibro-cartilagenous disks (3 mm diameter

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service