Skip to Content
Merck
All Photos(1)

Key Documents

239763-M

Sigma-Aldrich

Cycloheximide

Cycloheximide, CAS 66-81-9, is an antifungal antibiotic that inhibits protein synthesis in eukaryotes but not prokaryotes.

Synonym(s):

Cycloheximide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H23NO4
CAS Number:
Molecular Weight:
281.35
MDL number:
UNSPSC Code:
12352111
NACRES:
NA.77

Quality Level

Assay

≥90% (by assay)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white to brown

storage temp.

room temp

SMILES string

N1C(=O)CC(CC1=O)C[C@@H](O)[C@@H]2C[C@H](C[C@@H](C2=O)C)C

InChI

1S/C15H23NO4/c1-8-3-9(2)15(20)11(4-8)12(17)5-10-6-13(18)16-14(19)7-10/h8-12,17H,3-7H2,1-2H3,(H,16,18,19)/t8-,9-,11-,12+/m0/s1

InChI key

YPHMISFOHDHNIV-FSZOTQKASA-N

General description

An antifungal antibiotic that inhibits protein synthesis in eukaryotes but not in prokaryotes. Interacts directly with the translocase enzyme, interfering with the translocation step. Inhibits cell-free protein synthesis in eukaryotes. Competitively inhibits hFKBP12 (Ki = 3.4 µM). Triggers apoptosis in HL-60 cells, T-cell hybridomas, Burkitt′s lymphoma cells, and a variety of other cell types, including rodent macrophages. Induces DNA fragmentation in macrophages, however it inhibits DNA cleavage in rat thymocytes treated with thapsigargin, methylprednisolone, and ionomycin. Rapidly destroyed in alkaline solutions.
An antifungal antibiotic that inhibits protein synthesis in eukaryotes but not prokaryotes. Competitively inhibits hFKBP12 (Ki = 3.4 µM). Triggers apoptosis in HL-60 cells, T cell hybridomas, Burkitt’s lymphoma cells, and other cell types. A 100 mg/ml solution of Cycloheximide (Cat. No. 239765) in DMSO (sterile-filtered) is also available.

Warning

Toxicity: Highly Toxic & Carcinogenic / Teratogenic (I)

Other Notes

Christner, C., et al. 1999. J. Med. Chem.42, 3615.
Lu, Q. and Mellgreen,R.L. 1996. Arch. Biochem. Biophys.334, 175.
Chow, S.C., et al. 1995. Exp. Cell Res.216, 149.
Waring, P. 1990. J. Biol. Chem.265, 14476.
Obrig, T.G., et al. 1971. J. Biol. Chem.246, 174.
Pestka, S. 1971. Annu. Rev. Microbiol.25, 487.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Aquatic Chronic 2 - Muta. 2 - Repr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wenxin Zhang et al.
Acta pharmaceutica Sinica. B, 13(6), 2585-2600 (2023-07-10)
Mevalonate metabolism plays an important role in regulating tumor growth and progression; however, its role in immune evasion and immune checkpoint modulation remains unclear. Here, we found that non-small cell lung cancer (NSCLC) patients with higher plasma mevalonate response better
Ling Ding et al.
The Journal of clinical investigation, 133(1) (2023-01-04)
Understanding the regulatory mechanisms of PD-L1 expression in tumors provides key clues for improving immune checkpoint blockade efficacy or developing novel oncoimmunotherapy. Here, we showed that the FDA-approved sodium-glucose cotransporter-2 (SGLT2) inhibitor canagliflozin dramatically suppressed PD-L1 expression and enhanced T
Zhan Liu et al.
Cell death discovery, 10(1), 298-298 (2024-06-23)
LIMA1 is a LIM domain and Actin binding 1 protein that acts as a skeleton protein to promote cholesterol absorption, which makes it an ideal target for interfering with lipid metabolism. However, the detailed regulation of LIMA1 remains unclear. Here
Shifeng Li et al.
Theranostics, 10(4), 1719-1732 (2020-02-12)
The purpose of this study was to determine the effects of Kinesin family member 3A (KIF3A) on primary cilia and myofibroblast differentiation during silicosis by regulating Sonic hedgehog (SHH) signalling. Methods: Changes in primary cilia during silicosis and myofibroblast differentiation
Heidi M Blank et al.
EMBO reports, 24(9), e57372-e57372 (2023-07-27)
How cells coordinate their metabolism with division determines the rate of cell proliferation. Dynamic patterns of metabolite synthesis during the cell cycle are unexplored. We report the first isotope tracing analysis in synchronous, growing budding yeast cells. Synthesis of leucine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service