Skip to Content
Merck
All Photos(1)

Key Documents

405273

Sigma-Aldrich

INDY

Synonym(s):

INDY, (1Z)-1-(3-ethyl-5-hydroxy-2(3H)-benzothiazoylidene)-2-propanone, Dyrk/Clk Dual Inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H13NO2S
CAS Number:
Molecular Weight:
235.30
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 50 mg/mL

shipped in

wet ice

storage temp.

−20°C

General description

A benzothiazole compound that acts as an ATP competitive Dyrk inhibitor (IC50 = 0.24 µM and 0.23 µM for Dyrk1A and Dyrk1B, respectively), and displays >90% inhibition on DYRK2, DYRK3, CLK1, CLK4, casein kinase 1 (CSNK1D), and PIM1 among a panel of 66 kinases, in an in vitro screening assay. Unlike harmine, another potent Dyrk inhibitor, this compound does not affect MAOA activity. It is shown to decrease substrate phosphorylation of tau-protein at Thr212 in COS7 cells, dose-dependently, from 3 µM to 30 µM and ameliorates the inhibitory effect of Dyrk1A on NFAT response element mediated calcineurin/NFAT signaling in HEK293 cells. In addition, its prodrug, proINDY is shown to effectively reverse developmental defects associated with Dyrk1A overexpression in a Xenopus embryo model at 2.5 µM in vivo, without apparent toxicity.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Other Notes

Ogawa, Y., et al. 2010. Nat Commun1, 1.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service